Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44810348-44810595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD19
Entrez Gene ID
149478
Gene Name
BTB domain containing 19
Gene Aliases
-
UniGene
Hs.632400
Gene Chromosome Location
Chr.1: 44807442-44814199 on Build GRCh38
Transcripts
NM_001136537.1 XM_006710386.3 XM_011540803.2 XM_011540805.2 XM_017000447.1 XM_017000448.1 XM_017000449.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4882976 c.266T>C p.L89P Chr.1: 44810392-44810392 on Build GRCh38
1600037 c.292C>G p.R98G Chr.1: 44810418-44810418 on Build GRCh38
4482981 c.265C>T p.L89L Chr.1: 44810391-44810391 on Build GRCh38
5983739 c.258G>A p.L86L Chr.1: 44810384-44810384 on Build GRCh38
1197177 c.227C>G p.T76S Chr.1: 44810353-44810353 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372329975 Chr.1: 44810418-44810418 on Build GRCh38
rs376548632 Chr.1: 44810573-44810573 on Build GRCh38
rs377116613 Chr.1: 44810476-44810476 on Build GRCh38
rs146895380 Chr.1: 44810568-44810568 on Build GRCh38
rs886477079 Chr.1: 44810406-44810406 on Build GRCh38
rs150752523 Chr.1: 44810519-44810520 on Build GRCh38
rs770640373 Chr.1: 44810431-44810431 on Build GRCh38
rs369764765 Chr.1: 44810565-44810565 on Build GRCh38
rs749213058 Chr.1: 44810419-44810419 on Build GRCh38
rs1019641849 Chr.1: 44810421-44810421 on Build GRCh38
rs774219340 Chr.1: 44810461-44810461 on Build GRCh38
rs569604572 Chr.1: 44810489-44810489 on Build GRCh38
rs368300041 Chr.1: 44810378-44810378 on Build GRCh38
rs977887333 Chr.1: 44810404-44810404 on Build GRCh38
rs142560100 Chr.1: 44810488-44810488 on Build GRCh38
rs538241238 Chr.1: 44810493-44810493 on Build GRCh38
rs186821493 Chr.1: 44810574-44810574 on Build GRCh38
rs183942444 Chr.1: 44810570-44810570 on Build GRCh38

View Full Product Details