Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100140199-100140428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRMT13
Entrez Gene ID
54482
Gene Name
tRNA methyltransferase 13 homolog
Gene Aliases
CCDC76
UniGene
Hs.729095
Gene Chromosome Location
Chr.1: 100133080-100150498 on Build GRCh38
Transcripts
NM_019083.2 NR_135078.1 XM_005270945.3 XM_005270946.2 XM_017001504.1 XM_017001505.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5842861 c.409C>T p.L137F Chr.1: 100140422-100140422 on Build GRCh38
252420 c.395-2A>C p.? Chr.1: 100140406-100140406 on Build GRCh38
892937 c.385G>A p.A129T Chr.1: 100140242-100140242 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747080442 Chr.1: 100140389-100140389 on Build GRCh38
rs763559843 Chr.1: 100140306-100140309 on Build GRCh38
rs773499082 Chr.1: 100140388-100140388 on Build GRCh38
rs756883824 Chr.1: 100140267-100140267 on Build GRCh38
rs781269077 Chr.1: 100140271-100140271 on Build GRCh38
rs199548690 Chr.1: 100140362-100140362 on Build GRCh38
rs771068741 Chr.1: 100140395-100140395 on Build GRCh38
rs570284946 Chr.1: 100140386-100140386 on Build GRCh38
rs931080146 Chr.1: 100140264-100140264 on Build GRCh38
rs745840151 Chr.1: 100140281-100140281 on Build GRCh38
rs777094627 Chr.1: 100140401-100140401 on Build GRCh38
rs374711975 Chr.1: 100140261-100140261 on Build GRCh38
rs750972031 Chr.1: 100140266-100140266 on Build GRCh38
rs779664573 Chr.1: 100140359-100140359 on Build GRCh38
rs112436692 Chr.1: 100140268-100140268 on Build GRCh38
rs760089360 Chr.1: 100140402-100140402 on Build GRCh38
rs1000805184 Chr.1: 100140307-100140307 on Build GRCh38
rs755950827 Chr.1: 100140300-100140300 on Build GRCh38
rs757942895 Chr.1: 100140237-100140237 on Build GRCh38
rs761312216 Chr.1: 100140230-100140230 on Build GRCh38
rs750636958 Chr.1: 100140350-100140350 on Build GRCh38

View Full Product Details