Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 78243499-78243726 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP78
Entrez Gene ID
84131
Gene Name
centrosomal protein 78
Gene Aliases
C9orf81, IP63
UniGene
Hs.374421
Gene Chromosome Location
Chr.9: 78236062-78278431 on Build GRCh38
Transcripts
NM_001098802.1 NM_032171.1 XM_005252263.4 XM_005252265.2 XM_005252266.4 XM_017015192.1 XM_017015193.1 XM_017015194.1 XM_017015195.1 XM_017015196.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6352577 c.710A>G p.N237S Chr.9: 78243568-78243568 on Build GRCh38
7360952 c.724G>A p.D242N Chr.9: 78243582-78243582 on Build GRCh38
3908216 c.693T>C p.R231R Chr.9: 78243551-78243551 on Build GRCh38
3908213 c.650G>A p.R217H Chr.9: 78243508-78243508 on Build GRCh38
5953875 c.702G>C p.L234L Chr.9: 78243560-78243560 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs928120838 Chr.9: 78243590-78243590 on Build GRCh38
rs768319231 Chr.9: 78243543-78243543 on Build GRCh38
rs763435172 Chr.9: 78243584-78243584 on Build GRCh38
rs571801412 Chr.9: 78243673-78243673 on Build GRCh38
rs961163712 Chr.9: 78243605-78243605 on Build GRCh38
rs752835649 Chr.9: 78243632-78243632 on Build GRCh38
rs766852524 Chr.9: 78243606-78243606 on Build GRCh38
rs186712872 Chr.9: 78243568-78243568 on Build GRCh38
rs927687433 Chr.9: 78243658-78243658 on Build GRCh38
rs756222872 Chr.9: 78243639-78243639 on Build GRCh38
rs778124118 Chr.9: 78243665-78243665 on Build GRCh38
rs750169534 Chr.9: 78243679-78243679 on Build GRCh38
rs368402315 Chr.9: 78243538-78243538 on Build GRCh38
rs939138057 Chr.9: 78243693-78243693 on Build GRCh38
rs758316310 Chr.9: 78243685-78243685 on Build GRCh38
rs565154751 Chr.9: 78243531-78243531 on Build GRCh38
rs769494999 Chr.9: 78243558-78243558 on Build GRCh38
rs752102111 Chr.9: 78243607-78243607 on Build GRCh38
rs772131209 Chr.9: 78243529-78243529 on Build GRCh38
rs371528754 Chr.9: 78243612-78243612 on Build GRCh38
rs367644488 Chr.9: 78243684-78243684 on Build GRCh38
rs759575126 Chr.9: 78243608-78243609 on Build GRCh38
rs374919690 Chr.9: 78243657-78243657 on Build GRCh38
rs760069107 Chr.9: 78243611-78243611 on Build GRCh38
rs761319096 Chr.9: 78243550-78243550 on Build GRCh38
rs776232414 Chr.9: 78243549-78243549 on Build GRCh38

View Full Product Details