Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111980672-111980933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIXDC1
Entrez Gene ID
85458
Gene Name
DIX domain containing 1
Gene Aliases
CCD1
UniGene
Hs.655626
Gene Chromosome Location
Chr.11: 111927144-112022650 on Build GRCh38
Transcripts
NM_001037954.3 NM_033425.4 XM_017018466.1 XM_017018467.1 XM_017018468.1 XM_017018469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4017839 c.683G>A p.S228N Chr.11: 111980763-111980763 on Build GRCh38
4447884 c.752G>A p.G251E Chr.11: 111980832-111980832 on Build GRCh38
6292577 c.692G>T p.S231I Chr.11: 111980772-111980772 on Build GRCh38
6569033 c.718G>C p.E240Q Chr.11: 111980798-111980798 on Build GRCh38
5012433 c.725C>T p.A242V Chr.11: 111980805-111980805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781983847 Chr.11: 111980869-111980869 on Build GRCh38
rs781863884 Chr.11: 111980771-111980771 on Build GRCh38
rs782726158 Chr.11: 111980893-111980893 on Build GRCh38
rs782608370 Chr.11: 111980780-111980780 on Build GRCh38
rs782562272 Chr.11: 111980702-111980702 on Build GRCh38
rs782711106 Chr.11: 111980718-111980718 on Build GRCh38
rs782245932 Chr.11: 111980825-111980825 on Build GRCh38
rs587602283 Chr.11: 111980863-111980863 on Build GRCh38
rs587649456 Chr.11: 111980910-111980910 on Build GRCh38
rs782101240 Chr.11: 111980709-111980709 on Build GRCh38
rs782444294 Chr.11: 111980800-111980800 on Build GRCh38
rs782721787 Chr.11: 111980883-111980883 on Build GRCh38
rs782369905 Chr.11: 111980855-111980855 on Build GRCh38
rs782046560 Chr.11: 111980747-111980747 on Build GRCh38
rs781788899 Chr.11: 111980783-111980783 on Build GRCh38
rs782756299 Chr.11: 111980755-111980755 on Build GRCh38
rs782671817 Chr.11: 111980814-111980814 on Build GRCh38
rs782118665 Chr.11: 111980700-111980700 on Build GRCh38
rs587681755 Chr.11: 111980708-111980708 on Build GRCh38
rs782045971 Chr.11: 111980891-111980891 on Build GRCh38
rs587763740 Chr.11: 111980738-111980738 on Build GRCh38
rs781839060 Chr.11: 111980887-111980887 on Build GRCh38
rs145331507 Chr.11: 111980862-111980862 on Build GRCh38
rs782742979 Chr.11: 111980774-111980774 on Build GRCh38
rs782481530 Chr.11: 111980773-111980773 on Build GRCh38
rs782216264 Chr.11: 111980850-111980850 on Build GRCh38
rs782297945 Chr.11: 111980821-111980821 on Build GRCh38
rs782128262 Chr.11: 111980872-111980872 on Build GRCh38
rs781799796 Chr.11: 111980730-111980730 on Build GRCh38
rs111963828 Chr.11: 111980824-111980824 on Build GRCh38

View Full Product Details