Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7925197-7925446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTXN1
Entrez Gene ID
404217
Gene Name
cortexin 1
Gene Aliases
CTXN
UniGene
Hs.657978
Gene Chromosome Location
Chr.19: 7924496-7926166 on Build GRCh38
Transcripts
NM_206833.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
79709 c.141G>C p.V47V Chr.19: 7925398-7925398 on Build GRCh38
4653247 c.225G>C p.G75G Chr.19: 7925314-7925314 on Build GRCh38
4889419 c.212C>A p.A71E Chr.19: 7925327-7925327 on Build GRCh38
7126224 c.135G>C p.L45F Chr.19: 7925404-7925404 on Build GRCh38
6052131 c.142C>T p.R48C Chr.19: 7925397-7925397 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149989448 Chr.19: 7925415-7925415 on Build GRCh38
rs758866896 Chr.19: 7925275-7925275 on Build GRCh38
rs765602974 Chr.19: 7925372-7925372 on Build GRCh38
rs769584812 Chr.19: 7925314-7925314 on Build GRCh38
rs894438912 Chr.19: 7925258-7925258 on Build GRCh38
rs769483227 Chr.19: 7925243-7925243 on Build GRCh38
rs754481800 Chr.19: 7925406-7925406 on Build GRCh38
rs576794476 Chr.19: 7925269-7925269 on Build GRCh38
rs1002306758 Chr.19: 7925237-7925237 on Build GRCh38
rs376165674 Chr.19: 7925422-7925422 on Build GRCh38
rs758958911 Chr.19: 7925380-7925380 on Build GRCh38
rs767061441 Chr.19: 7925390-7925390 on Build GRCh38
rs770713330 Chr.19: 7925428-7925428 on Build GRCh38
rs752116910 Chr.19: 7925404-7925404 on Build GRCh38
rs780925269 Chr.19: 7925284-7925284 on Build GRCh38
rs747799569 Chr.19: 7925411-7925411 on Build GRCh38
rs779164740 Chr.19: 7925268-7925268 on Build GRCh38
rs780590547 Chr.19: 7925409-7925409 on Build GRCh38
rs764168000 Chr.19: 7925351-7925351 on Build GRCh38
rs1046581381 Chr.19: 7925228-7925228 on Build GRCh38
rs750801705 Chr.19: 7925377-7925377 on Build GRCh38
rs150061390 Chr.19: 7925270-7925270 on Build GRCh38
rs779688186 Chr.19: 7925277-7925277 on Build GRCh38
rs935804419 Chr.19: 7925232-7925232 on Build GRCh38
rs770909273 Chr.19: 7925324-7925324 on Build GRCh38
rs779740926 Chr.19: 7925239-7925239 on Build GRCh38
rs760678087 Chr.19: 7925340-7925340 on Build GRCh38
rs1027717321 Chr.19: 7925328-7925328 on Build GRCh38
rs767547661 Chr.19: 7925256-7925256 on Build GRCh38
rs936069640 Chr.19: 7925227-7925227 on Build GRCh38
rs761971083 Chr.19: 7925371-7925371 on Build GRCh38
rs775538876 Chr.19: 7925331-7925331 on Build GRCh38
rs754674096 Chr.19: 7925283-7925283 on Build GRCh38
rs747771660 Chr.19: 7925285-7925285 on Build GRCh38
rs906714925 Chr.19: 7925231-7925231 on Build GRCh38
rs776555839 Chr.19: 7925241-7925241 on Build GRCh38
rs760868375 Chr.19: 7925261-7925261 on Build GRCh38
rs761735401 Chr.19: 7925242-7925242 on Build GRCh38
rs762823580 Chr.19: 7925255-7925255 on Build GRCh38
rs888995572 Chr.19: 7925250-7925250 on Build GRCh38
rs752819371 Chr.19: 7925260-7925260 on Build GRCh38
rs868218155 Chr.19: 7925363-7925363 on Build GRCh38
rs1050223821 Chr.19: 7925249-7925250 on Build GRCh38
rs757581668 Chr.19: 7925267-7925267 on Build GRCh38
rs768472295 Chr.19: 7925240-7925240 on Build GRCh38
rs749221050 Chr.19: 7925323-7925323 on Build GRCh38
rs772915479 Chr.19: 7925317-7925317 on Build GRCh38
rs777606181 Chr.19: 7925419-7925419 on Build GRCh38
rs922975536 Chr.19: 7925311-7925311 on Build GRCh38
rs777678605 Chr.19: 7925214-7925214 on Build GRCh38
rs776861065 Chr.19: 7925353-7925353 on Build GRCh38
rs754007011 Chr.19: 7925266-7925266 on Build GRCh38
rs993579353 Chr.19: 7925324-7925325 on Build GRCh38
rs112578370 Chr.19: 7925249-7925249 on Build GRCh38
rs746575134 Chr.19: 7925278-7925278 on Build GRCh38

View Full Product Details