Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 38696569-38696796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf53
Entrez Gene ID
400359
Gene Name
chromosome 15 open reading frame 53
Gene Aliases
-
UniGene
Hs.448785
Gene Chromosome Location
Chr.15: 38696598-38700038 on Build GRCh38
Transcripts
NM_207444.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1372560 c.15G>T p.G5G Chr.15: 38696622-38696622 on Build GRCh38
3500716 c.25G>A p.D9N Chr.15: 38696632-38696632 on Build GRCh38
5769513 c.111C>G p.L37L Chr.15: 38696718-38696718 on Build GRCh38
3987915 c.117G>C p.A39A Chr.15: 38696724-38696724 on Build GRCh38
4054238 c.35T>C p.I12T Chr.15: 38696642-38696642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781059047 Chr.15: 38696617-38696617 on Build GRCh38
rs369659021 Chr.15: 38696776-38696776 on Build GRCh38
rs200650295 Chr.15: 38696593-38696593 on Build GRCh38
rs1056319970 Chr.15: 38696699-38696699 on Build GRCh38
rs745525527 Chr.15: 38696630-38696630 on Build GRCh38
rs7165988 Chr.15: 38696614-38696614 on Build GRCh38
rs764401573 Chr.15: 38696764-38696803 on Build GRCh38
rs374510527 Chr.15: 38696607-38696607 on Build GRCh38
rs865905617 Chr.15: 38696651-38696651 on Build GRCh38
rs769387175 Chr.15: 38696659-38696659 on Build GRCh38
rs747940079 Chr.15: 38696622-38696622 on Build GRCh38
rs146207678 Chr.15: 38696628-38696628 on Build GRCh38
rs760067243 Chr.15: 38696654-38696654 on Build GRCh38
rs756343414 Chr.15: 38696759-38696760 on Build GRCh38
rs150933904 Chr.15: 38696612-38696612 on Build GRCh38
rs147858205 Chr.15: 38696660-38696660 on Build GRCh38
rs777993486 Chr.15: 38696771-38696771 on Build GRCh38
rs142538711 Chr.15: 38696608-38696608 on Build GRCh38
rs760040814 Chr.15: 38696693-38696694 on Build GRCh38
rs753392863 Chr.15: 38696602-38696602 on Build GRCh38
rs993805091 Chr.15: 38696625-38696625 on Build GRCh38
rs778481261 Chr.15: 38696609-38696609 on Build GRCh38
rs955168480 Chr.15: 38696601-38696601 on Build GRCh38
rs769532560 Chr.15: 38696746-38696746 on Build GRCh38
rs11857596 Chr.15: 38696723-38696723 on Build GRCh38
rs753076817 Chr.15: 38696746-38696747 on Build GRCh38
rs767873269 Chr.15: 38696600-38696600 on Build GRCh38
rs771011689 Chr.15: 38696641-38696641 on Build GRCh38
rs757928043 Chr.15: 38696668-38696668 on Build GRCh38
rs758050913 Chr.15: 38696611-38696611 on Build GRCh38
rs935206105 Chr.15: 38696665-38696665 on Build GRCh38
rs767804676 Chr.15: 38696725-38696726 on Build GRCh38
rs761135849 Chr.15: 38696740-38696740 on Build GRCh38
rs750104040 Chr.15: 38696666-38696666 on Build GRCh38
rs767821377 Chr.15: 38696657-38696657 on Build GRCh38
rs559918753 Chr.15: 38696704-38696704 on Build GRCh38
rs747967690 Chr.15: 38696737-38696737 on Build GRCh38
rs755276979 Chr.15: 38696598-38696598 on Build GRCh38
rs773371984 Chr.15: 38696632-38696632 on Build GRCh38
rs763072414 Chr.15: 38696640-38696640 on Build GRCh38
rs146995135 Chr.15: 38696724-38696724 on Build GRCh38
rs774772177 Chr.15: 38696648-38696648 on Build GRCh38
rs764941959 Chr.15: 38696663-38696663 on Build GRCh38
rs751489520 Chr.15: 38696714-38696714 on Build GRCh38
rs946425899 Chr.15: 38696649-38696649 on Build GRCh38
rs1005910120 Chr.15: 38696669-38696669 on Build GRCh38
rs111493707 Chr.15: 38696774-38696774 on Build GRCh38

View Full Product Details