Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10091311-10091550 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf66
Entrez Gene ID
55337
Gene Name
chromosome 19 open reading frame 66
Gene Aliases
RyDEN
UniGene
Hs.655613
Gene Chromosome Location
Chr.19: 10086119-10093252 on Build GRCh38
Transcripts
NM_001308277.1 NM_018381.3 XM_011528121.1 XM_017026934.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6434607 c.491G>A p.G164D Chr.19: 10091478-10091478 on Build GRCh38
4615276 c.407_408insC p.R139fs*57 Chr.19: 10091547-10091548 on Build GRCh38
5668072 c.345A>C p.A115A Chr.19: 10091485-10091485 on Build GRCh38
4621521 c.360G>A p.P120P Chr.19: 10091500-10091500 on Build GRCh38
4620730 c.388G>A p.V130M Chr.19: 10091528-10091528 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756948185 Chr.19: 10091367-10091370 on Build GRCh38
rs756596403 Chr.19: 10091377-10091377 on Build GRCh38
rs902662036 Chr.19: 10091330-10091330 on Build GRCh38
rs746145785 Chr.19: 10091332-10091332 on Build GRCh38
rs780662691 Chr.19: 10091441-10091441 on Build GRCh38
rs752672390 Chr.19: 10091447-10091447 on Build GRCh38
rs754543483 Chr.19: 10091433-10091433 on Build GRCh38
rs1049393509 Chr.19: 10091518-10091518 on Build GRCh38
rs762466089 Chr.19: 10091528-10091528 on Build GRCh38
rs552291695 Chr.19: 10091414-10091414 on Build GRCh38
rs113702317 Chr.19: 10091408-10091408 on Build GRCh38
rs374119949 Chr.19: 10091500-10091500 on Build GRCh38
rs772326880 Chr.19: 10091333-10091333 on Build GRCh38
rs945225282 Chr.19: 10091468-10091468 on Build GRCh38
rs764031784 Chr.19: 10091353-10091353 on Build GRCh38
rs368721954 Chr.19: 10091376-10091376 on Build GRCh38
rs890068557 Chr.19: 10091467-10091467 on Build GRCh38
rs780801833 Chr.19: 10091370-10091371 on Build GRCh38
rs770590918 Chr.19: 10091488-10091488 on Build GRCh38
rs747175811 Chr.19: 10091501-10091501 on Build GRCh38
rs762374478 Chr.19: 10091369-10091369 on Build GRCh38
rs772961412 Chr.19: 10091328-10091328 on Build GRCh38
rs765022892 Chr.19: 10091415-10091415 on Build GRCh38
rs775507393 Chr.19: 10091340-10091340 on Build GRCh38
rs367703953 Chr.19: 10091512-10091512 on Build GRCh38
rs776842201 Chr.19: 10091524-10091524 on Build GRCh38
rs777326449 Chr.19: 10091361-10091361 on Build GRCh38
rs749123419 Chr.19: 10091471-10091471 on Build GRCh38
rs766062320 Chr.19: 10091380-10091380 on Build GRCh38
rs888144381 Chr.19: 10091519-10091519 on Build GRCh38
rs765759975 Chr.19: 10091372-10091372 on Build GRCh38
rs376197563 Chr.19: 10091373-10091373 on Build GRCh38
rs751135101 Chr.19: 10091390-10091390 on Build GRCh38
rs760838955 Chr.19: 10091342-10091342 on Build GRCh38
rs777605015 Chr.19: 10091451-10091451 on Build GRCh38
rs370225750 Chr.19: 10091448-10091448 on Build GRCh38

View Full Product Details