Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 127590934-127591181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARF5
Entrez Gene ID
381
Gene Name
ADP ribosylation factor 5
Gene Aliases
-
UniGene
Hs.653202
Gene Chromosome Location
Chr.7: 127588352-127591705 on Build GRCh38
Transcripts
NM_001662.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3674819 c.350G>T p.R117L Chr.7: 127590982-127590982 on Build GRCh38
7089644 c.331-1G>A p.? Chr.7: 127590962-127590962 on Build GRCh38
7000483 c.385G>A p.D129N Chr.7: 127591017-127591017 on Build GRCh38
1085149 c.349C>T p.R117W Chr.7: 127590981-127590981 on Build GRCh38
7265994 c.333G>A p.L111L Chr.7: 127590965-127590965 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745770296 Chr.7: 127591108-127591108 on Build GRCh38
rs770409712 Chr.7: 127591096-127591096 on Build GRCh38
rs781714849 Chr.7: 127590958-127590958 on Build GRCh38
rs1052657111 Chr.7: 127591124-127591124 on Build GRCh38
rs925226839 Chr.7: 127591120-127591120 on Build GRCh38
rs757346789 Chr.7: 127590955-127590955 on Build GRCh38
rs771831025 Chr.7: 127590995-127590995 on Build GRCh38
rs764976430 Chr.7: 127591037-127591037 on Build GRCh38
rs555345938 Chr.7: 127590954-127590954 on Build GRCh38
rs572157049 Chr.7: 127591088-127591088 on Build GRCh38
rs34457964 Chr.7: 127591073-127591073 on Build GRCh38
rs746372245 Chr.7: 127590959-127590959 on Build GRCh38
rs376366932 Chr.7: 127591055-127591055 on Build GRCh38
rs750899407 Chr.7: 127591083-127591083 on Build GRCh38
rs916226462 Chr.7: 127591080-127591080 on Build GRCh38
rs201108741 Chr.7: 127591029-127591029 on Build GRCh38
rs372711915 Chr.7: 127591131-127591131 on Build GRCh38
rs762721564 Chr.7: 127591128-127591128 on Build GRCh38
rs751785636 Chr.7: 127591077-127591077 on Build GRCh38
rs896833082 Chr.7: 127591058-127591058 on Build GRCh38
rs934046392 Chr.7: 127591142-127591142 on Build GRCh38
rs887907986 Chr.7: 127591010-127591010 on Build GRCh38
rs769687552 Chr.7: 127591111-127591111 on Build GRCh38
rs746526652 Chr.7: 127591022-127591022 on Build GRCh38
rs747749998 Chr.7: 127590991-127590991 on Build GRCh38
rs767496095 Chr.7: 127591078-127591078 on Build GRCh38
rs780593712 Chr.7: 127590982-127590982 on Build GRCh38
rs369276987 Chr.7: 127591067-127591067 on Build GRCh38
rs759398219 Chr.7: 127591035-127591035 on Build GRCh38
rs756706436 Chr.7: 127591084-127591084 on Build GRCh38
rs773043755 Chr.7: 127591005-127591005 on Build GRCh38
rs777471318 Chr.7: 127591094-127591094 on Build GRCh38
rs775471862 Chr.7: 127591113-127591113 on Build GRCh38
rs776036772 Chr.7: 127591100-127591100 on Build GRCh38
rs761969249 Chr.7: 127591138-127591138 on Build GRCh38
rs749643254 Chr.7: 127591087-127591087 on Build GRCh38
rs770409064 Chr.7: 127591028-127591028 on Build GRCh38
rs775287559 Chr.7: 127591043-127591043 on Build GRCh38
rs543068324 Chr.7: 127591044-127591044 on Build GRCh38
rs756432666 Chr.7: 127590974-127590974 on Build GRCh38
rs935178400 Chr.7: 127591123-127591123 on Build GRCh38
rs774562320 Chr.7: 127591136-127591136 on Build GRCh38

View Full Product Details