Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 2543169-2543442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRAT1
Entrez Gene ID
221927
Gene Name
BRCA1 associated ATM activator 1
Gene Aliases
BAAT1, C7orf27, RMFSL
UniGene
Hs.520623
Gene Chromosome Location
Chr.7: 2537810-2555758 on Build GRCh38
Transcripts
NM_152743.3 XM_005249643.4 XM_011515177.2 XM_011515178.1 XM_011515179.2 XM_011515181.2 XM_011515182.2 XM_011515183.1 XM_011515184.2 XM_011515186.2 XM_017011833.1 XM_017011834.1 XM_017011835.1 XM_017011836.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4964485 c.923+5G>A p.? Chr.7: 2543199-2543199 on Build GRCh38
7206278 c.870G>T p.L290L Chr.7: 2543257-2543257 on Build GRCh38
5026695 c.861G>A p.L287L Chr.7: 2543266-2543266 on Build GRCh38
3374598 c.810C>T p.P270P Chr.7: 2543317-2543317 on Build GRCh38
3026709 c.851C>T p.A284V Chr.7: 2543276-2543276 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752662283 Chr.7: 2543298-2543298 on Build GRCh38
rs560032715 Chr.7: 2543406-2543406 on Build GRCh38
rs755023265 Chr.7: 2543361-2543361 on Build GRCh38
rs752673566 Chr.7: 2543191-2543191 on Build GRCh38
rs77213198 Chr.7: 2543273-2543273 on Build GRCh38
rs76284903 Chr.7: 2543345-2543345 on Build GRCh38
rs773312644 Chr.7: 2543342-2543342 on Build GRCh38
rs758536859 Chr.7: 2543240-2543240 on Build GRCh38
rs766469928 Chr.7: 2543348-2543348 on Build GRCh38
rs940682856 Chr.7: 2543194-2543194 on Build GRCh38
rs573774447 Chr.7: 2543416-2543416 on Build GRCh38
rs1026084550 Chr.7: 2543381-2543381 on Build GRCh38
rs1014093234 Chr.7: 2543407-2543407 on Build GRCh38
rs766746960 Chr.7: 2543419-2543419 on Build GRCh38
rs374694299 Chr.7: 2543311-2543311 on Build GRCh38
rs767671540 Chr.7: 2543275-2543275 on Build GRCh38
rs963159318 Chr.7: 2543235-2543235 on Build GRCh38
rs746527927 Chr.7: 2543316-2543316 on Build GRCh38
rs151317339 Chr.7: 2543244-2543244 on Build GRCh38
rs750645922 Chr.7: 2543276-2543276 on Build GRCh38
rs980921384 Chr.7: 2543286-2543286 on Build GRCh38
rs200215583 Chr.7: 2543253-2543253 on Build GRCh38
rs201715454 Chr.7: 2543377-2543377 on Build GRCh38
rs748023062 Chr.7: 2543212-2543212 on Build GRCh38
rs751847390 Chr.7: 2543369-2543370 on Build GRCh38
rs781352986 Chr.7: 2543191-2543191 on Build GRCh38
rs372516307 Chr.7: 2543359-2543359 on Build GRCh38
rs113279845 Chr.7: 2543251-2543251 on Build GRCh38
rs374578110 Chr.7: 2543188-2543188 on Build GRCh38
rs367771169 Chr.7: 2543211-2543211 on Build GRCh38
rs575570226 Chr.7: 2543283-2543283 on Build GRCh38
rs767867860 Chr.7: 2543341-2543341 on Build GRCh38
rs761257350 Chr.7: 2543417-2543418 on Build GRCh38
rs758287252 Chr.7: 2543299-2543299 on Build GRCh38
rs745463379 Chr.7: 2543315-2543315 on Build GRCh38
rs769334756 Chr.7: 2543336-2543336 on Build GRCh38
rs371014140 Chr.7: 2543198-2543198 on Build GRCh38
rs140451075 Chr.7: 2543261-2543261 on Build GRCh38
rs751498648 Chr.7: 2543366-2543366 on Build GRCh38
rs775096485 Chr.7: 2543317-2543317 on Build GRCh38
rs934182833 Chr.7: 2543335-2543335 on Build GRCh38
rs570920491 Chr.7: 2543214-2543214 on Build GRCh38
rs764583480 Chr.7: 2543256-2543256 on Build GRCh38
rs756185597 Chr.7: 2543205-2543205 on Build GRCh38
rs201155141 Chr.7: 2543343-2543343 on Build GRCh38
rs909238271 Chr.7: 2543192-2543192 on Build GRCh38
rs1012610142 Chr.7: 2543344-2543344 on Build GRCh38
rs146546197 Chr.7: 2543301-2543301 on Build GRCh38
rs201793719 Chr.7: 2543340-2543340 on Build GRCh38
rs866641514 Chr.7: 2543225-2543225 on Build GRCh38
rs757013190 Chr.7: 2543368-2543368 on Build GRCh38
rs371711348 Chr.7: 2543305-2543305 on Build GRCh38
rs770541977 Chr.7: 2543221-2543221 on Build GRCh38
rs776318419 Chr.7: 2543237-2543237 on Build GRCh38
rs764087625 Chr.7: 2543362-2543362 on Build GRCh38
rs751831374 Chr.7: 2543187-2543187 on Build GRCh38
rs200455714 Chr.7: 2543319-2543319 on Build GRCh38
rs778075522 Chr.7: 2543293-2543294 on Build GRCh38
rs201041622 Chr.7: 2543243-2543243 on Build GRCh38
rs927524619 Chr.7: 2543292-2543292 on Build GRCh38
rs61742752 Chr.7: 2543218-2543218 on Build GRCh38
rs141320181 Chr.7: 2543302-2543302 on Build GRCh38
rs777504857 Chr.7: 2543217-2543217 on Build GRCh38
rs1053600024 Chr.7: 2543408-2543408 on Build GRCh38
rs778881636 Chr.7: 2543207-2543207 on Build GRCh38
rs750516424 Chr.7: 2543201-2543201 on Build GRCh38
rs567062198 Chr.7: 2543249-2543249 on Build GRCh38
rs766380023 Chr.7: 2543294-2543294 on Build GRCh38

View Full Product Details