Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140562570-140562795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APBB3
Entrez Gene ID
10307
Gene Name
amyloid beta precursor protein binding family B member 3
Gene Aliases
FE65L2, SRA
UniGene
Hs.529449
Gene Chromosome Location
Chr.5: 140558268-140564604 on Build GRCh38
Transcripts
NM_006051.3 NM_133172.2 NM_133173.2 NM_133174.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6601953 c.340C>T p.P114S Chr.5: 140562674-140562674 on Build GRCh38
3238936 c.337G>A p.E113K Chr.5: 140562677-140562677 on Build GRCh38
1061714 c.302C>A p.S101Y Chr.5: 140562712-140562712 on Build GRCh38
3946902 c.294C>G p.N98K Chr.5: 140562720-140562720 on Build GRCh38
482049 c.332G>A p.S111N Chr.5: 140562682-140562682 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748105061 Chr.5: 140562768-140562768 on Build GRCh38
rs544270502 Chr.5: 140562740-140562740 on Build GRCh38
rs34645592 Chr.5: 140562661-140562662 on Build GRCh38
rs761920886 Chr.5: 140562645-140562645 on Build GRCh38
rs776030408 Chr.5: 140562639-140562639 on Build GRCh38
rs907980946 Chr.5: 140562705-140562705 on Build GRCh38
rs978798910 Chr.5: 140562671-140562671 on Build GRCh38
rs768607693 Chr.5: 140562763-140562763 on Build GRCh38
rs778600820 Chr.5: 140562707-140562707 on Build GRCh38
rs746842390 Chr.5: 140562750-140562750 on Build GRCh38
rs767868295 Chr.5: 140562604-140562604 on Build GRCh38
rs756781817 Chr.5: 140562703-140562703 on Build GRCh38
rs751212712 Chr.5: 140562638-140562638 on Build GRCh38
rs757997881 Chr.5: 140562735-140562735 on Build GRCh38
rs140333079 Chr.5: 140562699-140562699 on Build GRCh38
rs960186735 Chr.5: 140562633-140562633 on Build GRCh38
rs200906807 Chr.5: 140562687-140562687 on Build GRCh38
rs766708790 Chr.5: 140562679-140562679 on Build GRCh38
rs555756503 Chr.5: 140562747-140562747 on Build GRCh38
rs146738104 Chr.5: 140562670-140562670 on Build GRCh38
rs953515100 Chr.5: 140562684-140562684 on Build GRCh38
rs767954170 Chr.5: 140562698-140562698 on Build GRCh38
rs567026684 Chr.5: 140562718-140562718 on Build GRCh38
rs760083930 Chr.5: 140562697-140562697 on Build GRCh38
rs773371197 Chr.5: 140562668-140562668 on Build GRCh38
rs756911355 Chr.5: 140562751-140562753 on Build GRCh38
rs1035899726 Chr.5: 140562628-140562628 on Build GRCh38
rs781155661 Chr.5: 140562764-140562764 on Build GRCh38
rs940529475 Chr.5: 140562706-140562706 on Build GRCh38
rs118128965 Chr.5: 140562734-140562734 on Build GRCh38
rs966077122 Chr.5: 140562752-140562752 on Build GRCh38
rs17118922 Chr.5: 140562656-140562656 on Build GRCh38
rs746902782 Chr.5: 140562630-140562630 on Build GRCh38
rs376996054 Chr.5: 140562767-140562767 on Build GRCh38
rs760993971 Chr.5: 140562641-140562641 on Build GRCh38
rs921917070 Chr.5: 140562675-140562675 on Build GRCh38

View Full Product Details