Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 178837212-178837439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC141
Entrez Gene ID
285025
Gene Name
coiled-coil domain containing 141
Gene Aliases
CAMDI
UniGene
Hs.324341
Gene Chromosome Location
Chr.2: 178825308-179050134 on Build GRCh38
Transcripts
NM_173648.3 XM_011510991.2 XM_011510992.2 XM_011510993.2 XM_017003867.1 XM_017003868.1 XM_017003869.1 XM_017003870.1 XM_017003871.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7094254 c.3815T>C p.F1272S Chr.2: 178837404-178837404 on Build GRCh38
5486205 c.3984T>C p.H1328H Chr.2: 178837235-178837235 on Build GRCh38
3425812 c.2146T>G p.Y716D Chr.2: 178837348-178837348 on Build GRCh38
5025804 c.3931_3932insACT p.A1311>DS Chr.2: 178837287-178837288 on Build GRCh38
3574587 c.3903C>T p.T1301T Chr.2: 178837316-178837316 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755074002 Chr.2: 178837370-178837370 on Build GRCh38
rs865980981 Chr.2: 178837308-178837308 on Build GRCh38
rs143659554 Chr.2: 178837271-178837271 on Build GRCh38
rs746060763 Chr.2: 178837277-178837277 on Build GRCh38
rs761715566 Chr.2: 178837303-178837303 on Build GRCh38
rs757632895 Chr.2: 178837253-178837253 on Build GRCh38
rs866690253 Chr.2: 178837387-178837387 on Build GRCh38
rs375789603 Chr.2: 178837352-178837352 on Build GRCh38
rs774155935 Chr.2: 178837318-178837318 on Build GRCh38
rs766077395 Chr.2: 178837315-178837315 on Build GRCh38
rs759017068 Chr.2: 178837322-178837322 on Build GRCh38
rs773303900 Chr.2: 178837393-178837393 on Build GRCh38
rs999988876 Chr.2: 178837292-178837292 on Build GRCh38
rs780183659 Chr.2: 178837356-178837356 on Build GRCh38
rs757184773 Chr.2: 178837337-178837338 on Build GRCh38
rs778099790 Chr.2: 178837385-178837387 on Build GRCh38
rs954693967 Chr.2: 178837282-178837282 on Build GRCh38
rs770340923 Chr.2: 178837414-178837414 on Build GRCh38
rs768622792 Chr.2: 178837299-178837299 on Build GRCh38
rs778886484 Chr.2: 178837355-178837356 on Build GRCh38
rs766999483 Chr.2: 178837336-178837336 on Build GRCh38
rs762871121 Chr.2: 178837312-178837312 on Build GRCh38
rs769814325 Chr.2: 178837389-178837389 on Build GRCh38
rs533080614 Chr.2: 178837351-178837351 on Build GRCh38
rs747054248 Chr.2: 178837298-178837298 on Build GRCh38
rs893602478 Chr.2: 178837323-178837323 on Build GRCh38
rs747263498 Chr.2: 178837358-178837358 on Build GRCh38
rs369418999 Chr.2: 178837340-178837340 on Build GRCh38
rs371380882 Chr.2: 178837283-178837283 on Build GRCh38
rs141212548 Chr.2: 178837278-178837278 on Build GRCh38
rs910795143 Chr.2: 178837295-178837295 on Build GRCh38
rs765769596 Chr.2: 178837345-178837345 on Build GRCh38
rs776628915 Chr.2: 178837300-178837300 on Build GRCh38
rs780280688 Chr.2: 178837281-178837281 on Build GRCh38
rs200623862 Chr.2: 178837397-178837397 on Build GRCh38
rs761543474 Chr.2: 178837400-178837400 on Build GRCh38
rs781177425 Chr.2: 178837372-178837372 on Build GRCh38
rs902581809 Chr.2: 178837288-178837288 on Build GRCh38
rs752784144 Chr.2: 178837246-178837248 on Build GRCh38
rs750741270 Chr.2: 178837347-178837347 on Build GRCh38
rs769688574 Chr.2: 178837304-178837304 on Build GRCh38
rs986137002 Chr.2: 178837286-178837286 on Build GRCh38
rs748086006 Chr.2: 178837374-178837374 on Build GRCh38
rs770849745 Chr.2: 178837401-178837401 on Build GRCh38
rs759166522 Chr.2: 178837419-178837419 on Build GRCh38

View Full Product Details