Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 76350953-76351182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC158
Entrez Gene ID
339965
Gene Name
coiled-coil domain containing 158
Gene Aliases
-
UniGene
Hs.529680
Gene Chromosome Location
Chr.4: 76313039-76421920 on Build GRCh38
Transcripts
NM_001042784.1 XM_005262974.4 XM_006714204.3 XM_011531908.2 XM_011531909.2 XM_011531910.2 XM_011531911.1 XM_011531912.2 XM_011531913.1 XM_011531914.2 XM_011531915.2 XM_011531916.2 XM_011531917.1 XM_017008160.1 XM_017008161.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4483005 c.2660C>T p.S887F Chr.4: 76351000-76351000 on Build GRCh38
4885462 c.2636C>T p.S879L Chr.4: 76351024-76351024 on Build GRCh38
1431084 c.2664+2T>C p.? Chr.4: 76350994-76350994 on Build GRCh38
1741864 c.2573C>T p.S858L Chr.4: 76351087-76351087 on Build GRCh38
6621778 c.2645C>T p.T882I Chr.4: 76351015-76351015 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775237376 Chr.4: 76351079-76351079 on Build GRCh38
rs758067497 Chr.4: 76350985-76350985 on Build GRCh38
rs748643136 Chr.4: 76351134-76351134 on Build GRCh38
rs1051909945 Chr.4: 76351152-76351152 on Build GRCh38
rs768857902 Chr.4: 76351099-76351099 on Build GRCh38
rs201737865 Chr.4: 76351085-76351085 on Build GRCh38
rs754956406 Chr.4: 76351129-76351129 on Build GRCh38
rs770692338 Chr.4: 76351059-76351059 on Build GRCh38
rs374260689 Chr.4: 76350987-76350987 on Build GRCh38
rs776345317 Chr.4: 76351062-76351062 on Build GRCh38
rs747372089 Chr.4: 76351146-76351146 on Build GRCh38
rs763839662 Chr.4: 76351094-76351094 on Build GRCh38
rs778339181 Chr.4: 76351145-76351145 on Build GRCh38
rs759046733 Chr.4: 76351075-76351075 on Build GRCh38
rs371657517 Chr.4: 76351007-76351007 on Build GRCh38
rs200617141 Chr.4: 76351153-76351153 on Build GRCh38
rs749358680 Chr.4: 76351023-76351023 on Build GRCh38
rs200146613 Chr.4: 76351035-76351035 on Build GRCh38
rs772275545 Chr.4: 76351043-76351043 on Build GRCh38
rs779497744 Chr.4: 76351131-76351131 on Build GRCh38
rs746706198 Chr.4: 76351057-76351057 on Build GRCh38
rs369910264 Chr.4: 76351116-76351116 on Build GRCh38
rs750608734 Chr.4: 76350996-76350996 on Build GRCh38
rs949235173 Chr.4: 76351049-76351049 on Build GRCh38
rs114770535 Chr.4: 76351077-76351077 on Build GRCh38
rs201598443 Chr.4: 76351004-76351004 on Build GRCh38
rs374874042 Chr.4: 76351026-76351026 on Build GRCh38
rs762841488 Chr.4: 76351084-76351084 on Build GRCh38
rs773595082 Chr.4: 76351048-76351048 on Build GRCh38
rs368649689 Chr.4: 76350999-76350999 on Build GRCh38
rs367607919 Chr.4: 76351037-76351037 on Build GRCh38
rs771655566 Chr.4: 76351145-76351145 on Build GRCh38
rs1052309085 Chr.4: 76351038-76351038 on Build GRCh38
rs377167162 Chr.4: 76351106-76351106 on Build GRCh38
rs199524814 Chr.4: 76351135-76351135 on Build GRCh38
rs756388872 Chr.4: 76351107-76351107 on Build GRCh38
rs766585104 Chr.4: 76351111-76351111 on Build GRCh38

View Full Product Details