Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 76968669-76968942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRHBP
Entrez Gene ID
1393
Gene Name
corticotropin releasing hormone binding protein
Gene Aliases
CRF-BP, CRFBP
UniGene
Hs.115617
Gene Chromosome Location
Chr.5: 76952713-76981143 on Build GRCh38
Transcripts
NM_001882.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4806704 c.847C>T p.R283C Chr.5: 76968763-76968763 on Build GRCh38
1671704 c.944G>T p.G315V Chr.5: 76968860-76968860 on Build GRCh38
1070268 c.870C>T p.H290H Chr.5: 76968786-76968786 on Build GRCh38
1070269 c.931G>T p.G311* Chr.5: 76968847-76968847 on Build GRCh38
4398697 c.902T>G p.L301R Chr.5: 76968818-76968818 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764188520 Chr.5: 76968766-76968766 on Build GRCh38
rs768911914 Chr.5: 76968882-76968882 on Build GRCh38
rs577642181 Chr.5: 76968824-76968824 on Build GRCh38
rs78871897 Chr.5: 76968755-76968755 on Build GRCh38
rs75290964 Chr.5: 76968739-76968739 on Build GRCh38
rs1007818776 Chr.5: 76968862-76968862 on Build GRCh38
rs766234801 Chr.5: 76968898-76968898 on Build GRCh38
rs377198989 Chr.5: 76968787-76968787 on Build GRCh38
rs75973668 Chr.5: 76968894-76968894 on Build GRCh38
rs778380094 Chr.5: 76968717-76968717 on Build GRCh38
rs76407422 Chr.5: 76968805-76968805 on Build GRCh38
rs753735786 Chr.5: 76968899-76968899 on Build GRCh38
rs77438825 Chr.5: 76968768-76968768 on Build GRCh38
rs778573715 Chr.5: 76968793-76968793 on Build GRCh38
rs553892661 Chr.5: 76968765-76968765 on Build GRCh38
rs775587058 Chr.5: 76968764-76968764 on Build GRCh38
rs777034631 Chr.5: 76968734-76968734 on Build GRCh38
rs757944609 Chr.5: 76968796-76968796 on Build GRCh38
rs77011613 Chr.5: 76968825-76968825 on Build GRCh38
rs771552936 Chr.5: 76968820-76968820 on Build GRCh38
rs76214989 Chr.5: 76968848-76968848 on Build GRCh38
rs753659305 Chr.5: 76968786-76968786 on Build GRCh38
rs186132339 Chr.5: 76968703-76968703 on Build GRCh38
rs200113770 Chr.5: 76968916-76968916 on Build GRCh38
rs372994637 Chr.5: 76968706-76968706 on Build GRCh38
rs774396648 Chr.5: 76968886-76968886 on Build GRCh38
rs79564270 Chr.5: 76968748-76968748 on Build GRCh38
rs771724310 Chr.5: 76968705-76968705 on Build GRCh38
rs780610415 Chr.5: 76968834-76968834 on Build GRCh38
rs769990720 Chr.5: 76968760-76968760 on Build GRCh38
rs78407170 Chr.5: 76968794-76968794 on Build GRCh38
rs966415593 Chr.5: 76968896-76968896 on Build GRCh38
rs777388217 Chr.5: 76968804-76968804 on Build GRCh38
rs368440034 Chr.5: 76968769-76968769 on Build GRCh38
rs151292463 Chr.5: 76968858-76968858 on Build GRCh38
rs766359445 Chr.5: 76968774-76968774 on Build GRCh38
rs866538801 Chr.5: 76968779-76968779 on Build GRCh38
rs759876702 Chr.5: 76968750-76968750 on Build GRCh38
rs140616064 Chr.5: 76968867-76968867 on Build GRCh38
rs767125968 Chr.5: 76968859-76968859 on Build GRCh38
rs563228859 Chr.5: 76968828-76968828 on Build GRCh38
rs746566970 Chr.5: 76968819-76968819 on Build GRCh38
rs771606098 Chr.5: 76968724-76968724 on Build GRCh38
rs775740166 Chr.5: 76968859-76968859 on Build GRCh38
rs747551950 Chr.5: 76968719-76968719 on Build GRCh38
rs760579941 Chr.5: 76968773-76968773 on Build GRCh38
rs542228751 Chr.5: 76968905-76968905 on Build GRCh38

View Full Product Details