Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 119146424-119146653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf30
Entrez Gene ID
152405
Gene Name
chromosome 3 open reading frame 30
Gene Aliases
TSCPA
UniGene
Hs.730099
Gene Chromosome Location
Chr.3: 119146150-119151455 on Build GRCh38
Transcripts
NM_152539.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
319088 c.374T>C p.V125A Chr.3: 119146563-119146563 on Build GRCh38
5702209 c.275_276CC>AA p.S92* Chr.3: 119146464-119146465 on Build GRCh38
1036675 c.396T>G p.G132G Chr.3: 119146585-119146585 on Build GRCh38
7093626 c.372T>C p.N124N Chr.3: 119146561-119146561 on Build GRCh38
4543944 c.346G>A p.G116S Chr.3: 119146535-119146535 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745810627 Chr.3: 119146461-119146461 on Build GRCh38
rs1052394631 Chr.3: 119146501-119146501 on Build GRCh38
rs777128841 Chr.3: 119146518-119146518 on Build GRCh38
rs374279095 Chr.3: 119146519-119146519 on Build GRCh38
rs765538779 Chr.3: 119146520-119146520 on Build GRCh38
rs780574210 Chr.3: 119146562-119146562 on Build GRCh38
rs112212634 Chr.3: 119146594-119146594 on Build GRCh38
rs746497808 Chr.3: 119146448-119146448 on Build GRCh38
rs762931267 Chr.3: 119146592-119146593 on Build GRCh38
rs776336581 Chr.3: 119146584-119146584 on Build GRCh38
rs751231587 Chr.3: 119146559-119146559 on Build GRCh38
rs770646159 Chr.3: 119146573-119146573 on Build GRCh38
rs138666071 Chr.3: 119146455-119146455 on Build GRCh38
rs771952122 Chr.3: 119146469-119146469 on Build GRCh38
rs772389838 Chr.3: 119146474-119146474 on Build GRCh38
rs759662566 Chr.3: 119146620-119146620 on Build GRCh38
rs777993427 Chr.3: 119146602-119146602 on Build GRCh38
rs1010490123 Chr.3: 119146510-119146510 on Build GRCh38
rs756738308 Chr.3: 119146561-119146561 on Build GRCh38
rs769648008 Chr.3: 119146462-119146462 on Build GRCh38
rs759179151 Chr.3: 119146616-119146616 on Build GRCh38
rs552664724 Chr.3: 119146611-119146611 on Build GRCh38
rs753040203 Chr.3: 119146523-119146523 on Build GRCh38
rs750348591 Chr.3: 119146563-119146563 on Build GRCh38
rs770928428 Chr.3: 119146511-119146511 on Build GRCh38
rs773087099 Chr.3: 119146517-119146517 on Build GRCh38
rs746755429 Chr.3: 119146505-119146505 on Build GRCh38
rs763798980 Chr.3: 119146535-119146535 on Build GRCh38
rs763348657 Chr.3: 119146532-119146532 on Build GRCh38
rs377325773 Chr.3: 119146447-119146447 on Build GRCh38
rs773369766 Chr.3: 119146479-119146479 on Build GRCh38
rs11550908 Chr.3: 119146485-119146485 on Build GRCh38
rs768389980 Chr.3: 119146599-119146599 on Build GRCh38
rs866723910 Chr.3: 119146499-119146499 on Build GRCh38
rs201847804 Chr.3: 119146603-119146603 on Build GRCh38
rs771000524 Chr.3: 119146605-119146605 on Build GRCh38
rs780178920 Chr.3: 119146575-119146575 on Build GRCh38

View Full Product Details