Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 45095825-45096088 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUOX2
Entrez Gene ID
50506
Gene Name
dual oxidase 2
Gene Aliases
LNOX2, NOXEF2, P138-TOX, TDH6, THOX2
UniGene
Hs.71377
Gene Chromosome Location
Chr.15: 45092653-45114161 on Build GRCh38
Transcripts
NM_014080.4 XM_005254421.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
386669 c.4015T>A p.W1339R Chr.15: 45095893-45095893 on Build GRCh38
4883019 c.4047C>A p.S1349S Chr.15: 45095861-45095861 on Build GRCh38
5849257 c.3848G>A p.G1283E Chr.15: 45096060-45096060 on Build GRCh38
4492285 c.3954C>T p.F1318F Chr.15: 45095954-45095954 on Build GRCh38
3887049 c.4046C>T p.S1349F Chr.15: 45095862-45095862 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771417585 Chr.15: 45096038-45096038 on Build GRCh38
rs762340753 Chr.15: 45095868-45095868 on Build GRCh38
rs368849023 Chr.15: 45095924-45095924 on Build GRCh38
rs913801510 Chr.15: 45096029-45096029 on Build GRCh38
rs776144364 Chr.15: 45095939-45095939 on Build GRCh38
rs888078705 Chr.15: 45095934-45095934 on Build GRCh38
rs768317705 Chr.15: 45095938-45095938 on Build GRCh38
rs751706037 Chr.15: 45095897-45095897 on Build GRCh38
rs766619891 Chr.15: 45095960-45095960 on Build GRCh38
rs759698581 Chr.15: 45095968-45095968 on Build GRCh38
rs778926241 Chr.15: 45095912-45095912 on Build GRCh38
rs528916465 Chr.15: 45095950-45095950 on Build GRCh38
rs755123065 Chr.15: 45095903-45095903 on Build GRCh38
rs990170176 Chr.15: 45096063-45096063 on Build GRCh38
rs367742457 Chr.15: 45096022-45096022 on Build GRCh38
rs756258652 Chr.15: 45095985-45095985 on Build GRCh38
rs145811119 Chr.15: 45096021-45096021 on Build GRCh38
rs375194680 Chr.15: 45095954-45095954 on Build GRCh38
rs750364813 Chr.15: 45095992-45095992 on Build GRCh38
rs140035596 Chr.15: 45095859-45095859 on Build GRCh38
rs913749110 Chr.15: 45095851-45095851 on Build GRCh38
rs779747989 Chr.15: 45095997-45095997 on Build GRCh38
rs780760868 Chr.15: 45096014-45096014 on Build GRCh38
rs768210021 Chr.15: 45096007-45096007 on Build GRCh38
rs777238983 Chr.15: 45095863-45095863 on Build GRCh38
rs1044930993 Chr.15: 45095901-45095901 on Build GRCh38
rs746613543 Chr.15: 45095928-45095928 on Build GRCh38
rs752828383 Chr.15: 45095975-45095975 on Build GRCh38
rs893622443 Chr.15: 45095889-45095889 on Build GRCh38
rs977450051 Chr.15: 45095857-45095857 on Build GRCh38
rs534696164 Chr.15: 45095998-45095998 on Build GRCh38
rs944676785 Chr.15: 45095852-45095852 on Build GRCh38
rs768030115 Chr.15: 45095862-45095862 on Build GRCh38
rs369478399 Chr.15: 45095993-45095993 on Build GRCh38
rs374795161 Chr.15: 45095921-45095921 on Build GRCh38
rs1036027188 Chr.15: 45095932-45095932 on Build GRCh38
rs147945181 Chr.15: 45095881-45095881 on Build GRCh38
rs148968706 Chr.15: 45096052-45096052 on Build GRCh38
rs775001939 Chr.15: 45095853-45095853 on Build GRCh38
rs751910050 Chr.15: 45095961-45095961 on Build GRCh38
rs781160106 Chr.15: 45095907-45095907 on Build GRCh38
rs550037603 Chr.15: 45095941-45095941 on Build GRCh38
rs774591147 Chr.15: 45096051-45096051 on Build GRCh38
rs749788680 Chr.15: 45096033-45096033 on Build GRCh38
rs200541410 Chr.15: 45095908-45095908 on Build GRCh38
rs200490615 Chr.15: 45095917-45095917 on Build GRCh38
rs988073471 Chr.15: 45096026-45096026 on Build GRCh38
rs763354648 Chr.15: 45095955-45095955 on Build GRCh38
rs766677894 Chr.15: 45095896-45095896 on Build GRCh38
rs966315712 Chr.15: 45096002-45096002 on Build GRCh38
rs767720307 Chr.15: 45095969-45095969 on Build GRCh38
rs149848729 Chr.15: 45095872-45095872 on Build GRCh38
rs564305753 Chr.15: 45095925-45095925 on Build GRCh38
rs947907970 Chr.15: 45095900-45095900 on Build GRCh38
rs548949995 Chr.15: 45095987-45095987 on Build GRCh38
rs1019582959 Chr.15: 45096001-45096001 on Build GRCh38
rs760616984 Chr.15: 45096027-45096027 on Build GRCh38
rs771855544 Chr.15: 45095847-45095847 on Build GRCh38
rs140110061 Chr.15: 45095883-45095883 on Build GRCh38
rs775841358 Chr.15: 45096064-45096064 on Build GRCh38
rs78020568 Chr.15: 45095940-45095940 on Build GRCh38
rs61730032 Chr.15: 45095942-45095942 on Build GRCh38
rs767698445 Chr.15: 45096053-45096053 on Build GRCh38

View Full Product Details