Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43042755-43043026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL7
Entrez Gene ID
9820
Gene Name
cullin 7
Gene Aliases
3M1, KIAA0076, dJ20C7.5
UniGene
Hs.520136
Gene Chromosome Location
Chr.6: 43037617-43053950 on Build GRCh38
Transcripts
NM_001168370.1 NM_014780.4 XM_005249503.2 XM_006715285.1 XM_011515019.2 XM_011515020.2 XM_011515021.1 XM_017011533.1 XM_017011534.1 XM_017011535.1 XM_017011536.1 XM_017011537.1 XM_017011538.1 XM_017011539.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
239486 c.3512G>A p.R1171H Chr.6: 43042935-43042935 on Build GRCh38
4650717 c.3743G>A p.R1248Q Chr.6: 43042956-43042956 on Build GRCh38
3394207 c.3588G>A p.A1196A Chr.6: 43042859-43042859 on Build GRCh38
7301474 c.3764G>A p.R1255H Chr.6: 43042935-43042935 on Build GRCh38
3874179 c.3490C>T p.R1164W Chr.6: 43042957-43042957 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754383585 Chr.6: 43042874-43042874 on Build GRCh38
rs764000209 Chr.6: 43042956-43042956 on Build GRCh38
rs776754610 Chr.6: 43042932-43042932 on Build GRCh38
rs959529000 Chr.6: 43042949-43042949 on Build GRCh38
rs867235173 Chr.6: 43042939-43042939 on Build GRCh38
rs902734461 Chr.6: 43042879-43042879 on Build GRCh38
rs915934070 Chr.6: 43042850-43042851 on Build GRCh38
rs1007218939 Chr.6: 43042791-43042791 on Build GRCh38
rs745751048 Chr.6: 43042935-43042935 on Build GRCh38
rs150591409 Chr.6: 43042868-43042868 on Build GRCh38
rs763687871 Chr.6: 43042852-43042852 on Build GRCh38
rs201742322 Chr.6: 43042844-43042844 on Build GRCh38
rs903674386 Chr.6: 43042873-43042873 on Build GRCh38
rs201981889 Chr.6: 43042838-43042838 on Build GRCh38
rs765683477 Chr.6: 43042991-43042991 on Build GRCh38
rs767078051 Chr.6: 43042980-43042980 on Build GRCh38
rs775318681 Chr.6: 43042944-43042944 on Build GRCh38
rs368840386 Chr.6: 43042891-43042891 on Build GRCh38
rs563011688 Chr.6: 43042923-43042923 on Build GRCh38
rs751032716 Chr.6: 43042860-43042860 on Build GRCh38
rs765006955 Chr.6: 43042820-43042820 on Build GRCh38
rs771044527 Chr.6: 43042777-43042777 on Build GRCh38
rs370419296 Chr.6: 43042863-43042863 on Build GRCh38
rs527664718 Chr.6: 43042994-43042994 on Build GRCh38
rs371526639 Chr.6: 43042997-43042997 on Build GRCh38
rs201515972 Chr.6: 43042836-43042836 on Build GRCh38
rs531667842 Chr.6: 43042936-43042936 on Build GRCh38
rs757487546 Chr.6: 43042999-43042999 on Build GRCh38
rs747268494 Chr.6: 43042916-43042916 on Build GRCh38
rs138726142 Chr.6: 43042899-43042899 on Build GRCh38
rs762601845 Chr.6: 43042842-43042842 on Build GRCh38
rs753293466 Chr.6: 43042993-43042993 on Build GRCh38
rs755500414 Chr.6: 43042984-43042984 on Build GRCh38
rs201135654 Chr.6: 43042957-43042957 on Build GRCh38
rs762681612 Chr.6: 43042946-43042946 on Build GRCh38
rs777832552 Chr.6: 43042910-43042910 on Build GRCh38
rs761625972 Chr.6: 43042965-43042965 on Build GRCh38
rs141610497 Chr.6: 43042859-43042859 on Build GRCh38
rs776515608 Chr.6: 43042778-43042778 on Build GRCh38
rs374772872 Chr.6: 43042865-43042865 on Build GRCh38
rs557518723 Chr.6: 43043002-43043002 on Build GRCh38
rs755554899 Chr.6: 43042878-43042878 on Build GRCh38
rs377501530 Chr.6: 43042802-43042802 on Build GRCh38
rs748490093 Chr.6: 43042898-43042898 on Build GRCh38
rs571347644 Chr.6: 43042983-43042983 on Build GRCh38

View Full Product Details