Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26412355-26412586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTN3A1
Entrez Gene ID
11119
Gene Name
butyrophilin subfamily 3 member A1
Gene Aliases
BT3.1, BTF5, BTN3.1, CD277
UniGene
Hs.191510
Gene Chromosome Location
Chr.6: 26402237-26415216 on Build GRCh38
Transcripts
NM_001145008.1 NM_001145009.1 NM_007048.5 NM_194441.2 XM_005248833.2 XM_005248834.3 XM_006714983.2 XM_011514268.2

Gene Symbol
BTN3A2
Entrez Gene ID
11118
Gene Name
butyrophilin subfamily 3 member A2
Gene Aliases
BT3.2, BTF4, BTN3.2, CD277
UniGene
Hs.376046
Gene Chromosome Location
Chr.6: 26365159-26451697 on Build GRCh38
Transcripts
NM_001145008.1 NM_001145009.1 NM_007048.5 NM_194441.2 XM_005248833.2 XM_005248834.3 XM_006714983.2 XM_011514268.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6502691 c.1127G>C p.G376A Chr.6: 26412520-26412520 on Build GRCh38
5988183 c.1120G>T p.E374* Chr.6: 26412513-26412513 on Build GRCh38
6279346 c.1019-8C>T p.? Chr.6: 26412404-26412404 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1001901395 Chr.6: 26412447-26412447 on Build GRCh38
rs775795385 Chr.6: 26412534-26412534 on Build GRCh38
rs893369541 Chr.6: 26412506-26412506 on Build GRCh38
rs778449920 Chr.6: 26412472-26412472 on Build GRCh38
rs1045814375 Chr.6: 26412378-26412378 on Build GRCh38
rs537547277 Chr.6: 26412490-26412490 on Build GRCh38
rs926008084 Chr.6: 26412400-26412400 on Build GRCh38
rs186430588 Chr.6: 26412477-26412477 on Build GRCh38
rs1053212416 Chr.6: 26412426-26412426 on Build GRCh38
rs181798750 Chr.6: 26412414-26412414 on Build GRCh38
rs967129630 Chr.6: 26412563-26412563 on Build GRCh38
rs757983023 Chr.6: 26412493-26412493 on Build GRCh38
rs763327535 Chr.6: 26412550-26412550 on Build GRCh38
rs558114756 Chr.6: 26412462-26412462 on Build GRCh38
rs1020391255 Chr.6: 26412561-26412561 on Build GRCh38
rs746488191 Chr.6: 26412460-26412460 on Build GRCh38

View Full Product Details