Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144354016-144354281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM249
Entrez Gene ID
340393
Gene Name
transmembrane protein 249
Gene Aliases
C8ORFK29
UniGene
Hs.122231
Gene Chromosome Location
Chr.8: 144353226-144355609 on Build GRCh38
Transcripts
NM_001252402.2 NM_001252404.2 NM_001280561.1 NR_047684.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4162843 c.371C>A p.S124Y Chr.8: 144354025-144354025 on Build GRCh38
5717468 c.231C>T p.F77F Chr.8: 144354252-144354252 on Build GRCh38
4957171 c.318G>A p.S106S Chr.8: 144354078-144354078 on Build GRCh38
4956325 c.354C>T p.R118R Chr.8: 144354042-144354042 on Build GRCh38
4953473 c.212G>T p.S71I Chr.8: 144354271-144354271 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782689481 Chr.8: 144354158-144354158 on Build GRCh38
rs782425295 Chr.8: 144354042-144354042 on Build GRCh38
rs782357115 Chr.8: 144354177-144354177 on Build GRCh38
rs782782778 Chr.8: 144354187-144354187 on Build GRCh38
rs925591824 Chr.8: 144354072-144354072 on Build GRCh38
rs1025087178 Chr.8: 144354208-144354208 on Build GRCh38
rs782198999 Chr.8: 144354056-144354056 on Build GRCh38
rs782773021 Chr.8: 144354117-144354117 on Build GRCh38
rs781871639 Chr.8: 144354157-144354157 on Build GRCh38
rs113558963 Chr.8: 144354147-144354147 on Build GRCh38
rs782112141 Chr.8: 144354113-144354113 on Build GRCh38
rs116055973 Chr.8: 144354239-144354239 on Build GRCh38
rs781960407 Chr.8: 144354090-144354090 on Build GRCh38
rs781924578 Chr.8: 144354045-144354045 on Build GRCh38
rs880701 Chr.8: 144354164-144354164 on Build GRCh38
rs781983211 Chr.8: 144354181-144354181 on Build GRCh38
rs981168055 Chr.8: 144354071-144354071 on Build GRCh38
rs782219135 Chr.8: 144354163-144354163 on Build GRCh38
rs782017518 Chr.8: 144354126-144354126 on Build GRCh38
rs880702 Chr.8: 144354169-144354169 on Build GRCh38
rs905268594 Chr.8: 144354219-144354219 on Build GRCh38
rs781803109 Chr.8: 144354137-144354137 on Build GRCh38
rs782697417 Chr.8: 144354131-144354131 on Build GRCh38
rs782710592 Chr.8: 144354190-144354190 on Build GRCh38
rs1036882452 Chr.8: 144354188-144354188 on Build GRCh38
rs782334659 Chr.8: 144354060-144354060 on Build GRCh38
rs944157656 Chr.8: 144354123-144354123 on Build GRCh38
rs782127394 Chr.8: 144354185-144354185 on Build GRCh38
rs781817170 Chr.8: 144354203-144354203 on Build GRCh38
rs969439577 Chr.8: 144354070-144354070 on Build GRCh38
rs781875435 Chr.8: 144354250-144354250 on Build GRCh38
rs374933177 Chr.8: 144354119-144354119 on Build GRCh38
rs1032759535 Chr.8: 144354261-144354261 on Build GRCh38
rs781903181 Chr.8: 144354189-144354189 on Build GRCh38
rs782601984 Chr.8: 144354162-144354162 on Build GRCh38
rs782460376 Chr.8: 144354160-144354160 on Build GRCh38
rs540720898 Chr.8: 144354133-144354133 on Build GRCh38
rs782480458 Chr.8: 144354154-144354154 on Build GRCh38
rs782495803 Chr.8: 144354210-144354210 on Build GRCh38
rs782164438 Chr.8: 144354128-144354128 on Build GRCh38
rs1014486920 Chr.8: 144354202-144354202 on Build GRCh38
rs958273325 Chr.8: 144354073-144354073 on Build GRCh38

View Full Product Details