Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40845191-40845416 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2A6
Entrez Gene ID
1548
Gene Name
cytochrome P450 family 2 subfamily A member 6
Gene Aliases
CPA6, CYP2A, CYP2A3, CYPIIA6, P450C2A, P450PB
UniGene
Hs.250615
Gene Chromosome Location
Chr.19: 40843538-40850447 on Build GRCh38
Transcripts
NM_000762.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4985329 c.1093G>A p.V365M Chr.19: 40845362-40845362 on Build GRCh38
6888748 c.1098C>T p.I366I Chr.19: 40845357-40845357 on Build GRCh38
7129703 c.1149C>T p.F383F Chr.19: 40845306-40845306 on Build GRCh38
439563 c.1091A>C p.D364A Chr.19: 40845364-40845364 on Build GRCh38
712828 c.1103T>C p.M368T Chr.19: 40845352-40845352 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369136003 Chr.19: 40845302-40845302 on Build GRCh38
rs879816581 Chr.19: 40845232-40845232 on Build GRCh38
rs751304482 Chr.19: 40845359-40845359 on Build GRCh38
rs752841802 Chr.19: 40845363-40845363 on Build GRCh38
rs201588233 Chr.19: 40845290-40845290 on Build GRCh38
rs142724678 Chr.19: 40845293-40845293 on Build GRCh38
rs751427594 Chr.19: 40845304-40845304 on Build GRCh38
rs202136237 Chr.19: 40845284-40845284 on Build GRCh38
rs377119322 Chr.19: 40845249-40845249 on Build GRCh38
rs771265125 Chr.19: 40845333-40845333 on Build GRCh38
rs114558780 Chr.19: 40845322-40845322 on Build GRCh38
rs372636737 Chr.19: 40845311-40845311 on Build GRCh38
rs72547582 Chr.19: 40845295-40845295 on Build GRCh38
rs1015811111 Chr.19: 40845296-40845297 on Build GRCh38
rs185304177 Chr.19: 40845321-40845321 on Build GRCh38
rs778129171 Chr.19: 40845274-40845274 on Build GRCh38
rs28399455 Chr.19: 40845285-40845285 on Build GRCh38
rs766817886 Chr.19: 40845222-40845222 on Build GRCh38
rs757428419 Chr.19: 40845383-40845383 on Build GRCh38
rs145036049 Chr.19: 40845340-40845340 on Build GRCh38
rs375925239 Chr.19: 40845384-40845384 on Build GRCh38
rs386809288 Chr.19: 40845264-40845265 on Build GRCh38
rs923853269 Chr.19: 40845387-40845387 on Build GRCh38
rs775124075 Chr.19: 40845229-40845229 on Build GRCh38
rs532833480 Chr.19: 40845273-40845273 on Build GRCh38
rs756390851 Chr.19: 40845313-40845313 on Build GRCh38
rs771496020 Chr.19: 40845281-40845281 on Build GRCh38
rs754083708 Chr.19: 40845381-40845381 on Build GRCh38
rs761393328 Chr.19: 40845345-40845345 on Build GRCh38
rs576392235 Chr.19: 40845242-40845242 on Build GRCh38
rs4803380 Chr.19: 40845264-40845264 on Build GRCh38
rs774691324 Chr.19: 40845283-40845283 on Build GRCh38
rs778019189 Chr.19: 40845370-40845370 on Build GRCh38
rs142447929 Chr.19: 40845346-40845346 on Build GRCh38
rs779290232 Chr.19: 40845336-40845336 on Build GRCh38
rs749640469 Chr.19: 40845276-40845276 on Build GRCh38
rs758423225 Chr.19: 40845263-40845263 on Build GRCh38
rs772964366 Chr.19: 40845351-40845351 on Build GRCh38
rs756883463 Chr.19: 40845259-40845259 on Build GRCh38
rs781140086 Chr.19: 40845309-40845309 on Build GRCh38
rs763849009 Chr.19: 40845253-40845253 on Build GRCh38
rs756155939 Chr.19: 40845365-40845365 on Build GRCh38
rs766120898 Chr.19: 40845358-40845358 on Build GRCh38
rs961941376 Chr.19: 40845300-40845300 on Build GRCh38
rs28399454 Chr.19: 40845362-40845362 on Build GRCh38
rs780602964 Chr.19: 40845395-40845395 on Build GRCh38
rs759408460 Chr.19: 40845360-40845360 on Build GRCh38
rs369798042 Chr.19: 40845261-40845261 on Build GRCh38
rs1029356350 Chr.19: 40845338-40845338 on Build GRCh38
rs762887319 Chr.19: 40845352-40845352 on Build GRCh38
rs150586234 Chr.19: 40845306-40845306 on Build GRCh38
rs759381042 Chr.19: 40845279-40845279 on Build GRCh38
rs764924638 Chr.19: 40845288-40845288 on Build GRCh38
rs781286490 Chr.19: 40845272-40845272 on Build GRCh38
rs758887029 Chr.19: 40845386-40845386 on Build GRCh38
rs776018244 Chr.19: 40845341-40845341 on Build GRCh38
rs59992959 Chr.19: 40845339-40845339 on Build GRCh38
rs147883801 Chr.19: 40845356-40845356 on Build GRCh38
rs60286635 Chr.19: 40845265-40845265 on Build GRCh38

View Full Product Details