Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32211133-32211406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCDC2B
Entrez Gene ID
149069
Gene Name
doublecortin domain containing 2B
Gene Aliases
-
UniGene
Hs.424588
Gene Chromosome Location
Chr.1: 32208813-32216196 on Build GRCh38
Transcripts
NM_001099434.1 XM_017000413.1 XM_017000414.1 XM_011541795.2 XM_017001814.1 XM_017001815.1 XM_017001817.1 XM_017001818.1 XM_017001819.1 XM_017001821.1 XM_017001827.1 XM_017001828.1

Gene Symbol
TMEM234
Entrez Gene ID
56063
Gene Name
transmembrane protein 234
Gene Aliases
AASL548, C1orf91, PRO1105, RP4-622L5, dJ622L5.7
UniGene
Hs.272299
Gene Chromosome Location
Chr.1: 32204807-32222371 on Build GRCh38
Transcripts
NM_001099434.1 XM_017000413.1 XM_017000414.1 XM_011541795.2 XM_017001814.1 XM_017001815.1 XM_017001817.1 XM_017001818.1 XM_017001819.1 XM_017001821.1 XM_017001827.1 XM_017001828.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3488110 c.284G>A p.G95E Chr.1: 32211289-32211289 on Build GRCh38
7363799 c.293C>A p.P98Q Chr.1: 32211298-32211298 on Build GRCh38
3689606 c.267-1G>A p.? Chr.1: 32211271-32211271 on Build GRCh38
7363800 c.267C>T p.H89H Chr.1: 32211272-32211272 on Build GRCh38
6640700 c.300G>A p.G100G Chr.1: 32211305-32211305 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976196483 Chr.1: 32211222-32211222 on Build GRCh38
rs759105551 Chr.1: 32211326-32211327 on Build GRCh38
rs761475363 Chr.1: 32211358-32211358 on Build GRCh38
rs377409613 Chr.1: 32211370-32211370 on Build GRCh38
rs771716694 Chr.1: 32211354-32211355 on Build GRCh38
rs750162862 Chr.1: 32211369-32211369 on Build GRCh38
rs572238102 Chr.1: 32211333-32211333 on Build GRCh38
rs779280222 Chr.1: 32211316-32211316 on Build GRCh38
rs138604564 Chr.1: 32211221-32211221 on Build GRCh38
rs760154954 Chr.1: 32211336-32211336 on Build GRCh38
rs990755822 Chr.1: 32211199-32211199 on Build GRCh38
rs549233990 Chr.1: 32211220-32211220 on Build GRCh38
rs375619141 Chr.1: 32211263-32211263 on Build GRCh38
rs566576562 Chr.1: 32211187-32211187 on Build GRCh38
rs931987471 Chr.1: 32211237-32211239 on Build GRCh38
rs538191171 Chr.1: 32211232-32211232 on Build GRCh38
rs539040584 Chr.1: 32211321-32211321 on Build GRCh38
rs188979173 Chr.1: 32211190-32211190 on Build GRCh38
rs1045365479 Chr.1: 32211264-32211264 on Build GRCh38
rs777830542 Chr.1: 32211310-32211310 on Build GRCh38
rs909977143 Chr.1: 32211189-32211189 on Build GRCh38
rs750204042 Chr.1: 32211270-32211270 on Build GRCh38
rs966946267 Chr.1: 32211383-32211383 on Build GRCh38
rs141330031 Chr.1: 32211272-32211272 on Build GRCh38
rs752813964 Chr.1: 32211294-32211294 on Build GRCh38
rs764806766 Chr.1: 32211363-32211363 on Build GRCh38
rs372581183 Chr.1: 32211259-32211259 on Build GRCh38
rs765040861 Chr.1: 32211266-32211266 on Build GRCh38
rs780142043 Chr.1: 32211283-32211283 on Build GRCh38
rs949999383 Chr.1: 32211250-32211250 on Build GRCh38
rs941192967 Chr.1: 32211296-32211296 on Build GRCh38
rs768252289 Chr.1: 32211345-32211345 on Build GRCh38
rs200739008 Chr.1: 32211332-32211332 on Build GRCh38
rs889735568 Chr.1: 32211284-32211284 on Build GRCh38
rs376478983 Chr.1: 32211213-32211213 on Build GRCh38
rs201678864 Chr.1: 32211308-32211308 on Build GRCh38
rs570013445 Chr.1: 32211367-32211369 on Build GRCh38
rs776036715 Chr.1: 32211348-32211348 on Build GRCh38
rs374509093 Chr.1: 32211366-32211366 on Build GRCh38
rs1001210066 Chr.1: 32211278-32211278 on Build GRCh38
rs749490106 Chr.1: 32211313-32211313 on Build GRCh38
rs761480603 Chr.1: 32211261-32211261 on Build GRCh38
rs773967365 Chr.1: 32211272-32211273 on Build GRCh38
rs760246898 Chr.1: 32211239-32211239 on Build GRCh38
rs771081630 Chr.1: 32211314-32211314 on Build GRCh38
rs753175032 Chr.1: 32211181-32211181 on Build GRCh38
rs923237488 Chr.1: 32211227-32211227 on Build GRCh38
rs781225463 Chr.1: 32211174-32211174 on Build GRCh38
rs763721557 Chr.1: 32211258-32211258 on Build GRCh38
rs892898819 Chr.1: 32211289-32211289 on Build GRCh38
rs1054577600 Chr.1: 32211279-32211279 on Build GRCh38

View Full Product Details