Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8042684-8042933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX15B
Entrez Gene ID
247
Gene Name
arachidonate 15-lipoxygenase, type B
Gene Aliases
15-LOX-2
UniGene
Hs.111256
Gene Chromosome Location
Chr.17: 8039040-8049134 on Build GRCh38
Transcripts
NM_001039130.1 NM_001039131.1 NM_001141.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6339733 c.608G>A p.R203H Chr.17: 8042816-8042816 on Build GRCh38
7308704 c.573-8C>T p.? Chr.17: 8042773-8042773 on Build GRCh38
1387566 c.613G>T p.G205W Chr.17: 8042821-8042821 on Build GRCh38
4639006 c.670G>A p.A224T Chr.17: 8042878-8042878 on Build GRCh38
5892311 c.644G>A p.R215K Chr.17: 8042852-8042852 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373709924 Chr.17: 8042814-8042814 on Build GRCh38
rs918170790 Chr.17: 8042763-8042763 on Build GRCh38
rs777368790 Chr.17: 8042808-8042808 on Build GRCh38
rs1054144873 Chr.17: 8042899-8042899 on Build GRCh38
rs751250458 Chr.17: 8042902-8042902 on Build GRCh38
rs1018162859 Chr.17: 8042733-8042733 on Build GRCh38
rs868821090 Chr.17: 8042788-8042788 on Build GRCh38
rs867856560 Chr.17: 8042786-8042786 on Build GRCh38
rs765779158 Chr.17: 8042860-8042860 on Build GRCh38
rs921961228 Chr.17: 8042704-8042704 on Build GRCh38
rs779778522 Chr.17: 8042732-8042732 on Build GRCh38
rs375265452 Chr.17: 8042816-8042816 on Build GRCh38
rs981183333 Chr.17: 8042892-8042892 on Build GRCh38
rs76872015 Chr.17: 8042714-8042714 on Build GRCh38
rs750713821 Chr.17: 8042861-8042861 on Build GRCh38
rs575698344 Chr.17: 8042756-8042756 on Build GRCh38
rs949699816 Chr.17: 8042791-8042791 on Build GRCh38
rs890929125 Chr.17: 8042723-8042723 on Build GRCh38
rs775520681 Chr.17: 8042711-8042711 on Build GRCh38
rs763925528 Chr.17: 8042867-8042867 on Build GRCh38
rs768715727 Chr.17: 8042824-8042824 on Build GRCh38
rs758982554 Chr.17: 8042895-8042895 on Build GRCh38
rs780918756 Chr.17: 8042759-8042759 on Build GRCh38
rs565518460 Chr.17: 8042778-8042778 on Build GRCh38
rs776748473 Chr.17: 8042828-8042828 on Build GRCh38
rs563680504 Chr.17: 8042715-8042715 on Build GRCh38
rs905548726 Chr.17: 8042830-8042830 on Build GRCh38
rs762250775 Chr.17: 8042841-8042841 on Build GRCh38
rs772132269 Chr.17: 8042815-8042815 on Build GRCh38
rs769869330 Chr.17: 8042790-8042790 on Build GRCh38
rs867766950 Chr.17: 8042832-8042832 on Build GRCh38
rs748087577 Chr.17: 8042764-8042764 on Build GRCh38
rs962614879 Chr.17: 8042751-8042751 on Build GRCh38
rs370631978 Chr.17: 8042757-8042757 on Build GRCh38
rs766693592 Chr.17: 8042896-8042896 on Build GRCh38

View Full Product Details