Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29331702-29331969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1B1
Entrez Gene ID
162
Gene Name
adaptor related protein complex 1 beta 1 subunit
Gene Aliases
ADTB1, AP105A, BAM22, CLAPB2
UniGene
Hs.368794
Gene Chromosome Location
Chr.22: 29327680-29388583 on Build GRCh38
Transcripts
NM_001127.3 NM_001166019.1 NM_145730.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2936396 c.2436C>T p.L812L Chr.22: 29331790-29331790 on Build GRCh38
1484141 c.2406G>A p.S802S Chr.22: 29331820-29331820 on Build GRCh38
5859129 c.2360C>T p.S787F Chr.22: 29331845-29331845 on Build GRCh38
4854850 c.2397G>A p.T799T Chr.22: 29331829-29331829 on Build GRCh38
3553147 c.2302C>T p.P768S Chr.22: 29331903-29331903 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766392381 Chr.22: 29331912-29331912 on Build GRCh38
rs750563769 Chr.22: 29331839-29331839 on Build GRCh38
rs2857465 Chr.22: 29331897-29331897 on Build GRCh38
rs546260476 Chr.22: 29331879-29331879 on Build GRCh38
rs937210340 Chr.22: 29331905-29331905 on Build GRCh38
rs200642829 Chr.22: 29331738-29331738 on Build GRCh38
rs1044496543 Chr.22: 29331812-29331812 on Build GRCh38
rs753059593 Chr.22: 29331922-29331922 on Build GRCh38
rs371753455 Chr.22: 29331793-29331793 on Build GRCh38
rs1055622652 Chr.22: 29331939-29331939 on Build GRCh38
rs755475534 Chr.22: 29331920-29331920 on Build GRCh38
rs922218153 Chr.22: 29331824-29331824 on Build GRCh38
rs138608403 Chr.22: 29331892-29331892 on Build GRCh38
rs552463610 Chr.22: 29331758-29331758 on Build GRCh38
rs144915223 Chr.22: 29331740-29331740 on Build GRCh38
rs535159123 Chr.22: 29331887-29331887 on Build GRCh38
rs1035639687 Chr.22: 29331851-29331851 on Build GRCh38
rs369442816 Chr.22: 29331863-29331863 on Build GRCh38
rs780548317 Chr.22: 29331852-29331852 on Build GRCh38
rs367968927 Chr.22: 29331747-29331747 on Build GRCh38
rs375276977 Chr.22: 29331778-29331778 on Build GRCh38
rs774062307 Chr.22: 29331739-29331739 on Build GRCh38
rs749731994 Chr.22: 29331737-29331737 on Build GRCh38
rs746601818 Chr.22: 29331940-29331940 on Build GRCh38
rs754396935 Chr.22: 29331914-29331914 on Build GRCh38
rs174765 Chr.22: 29331877-29331877 on Build GRCh38
rs905931994 Chr.22: 29331832-29331832 on Build GRCh38
rs974921588 Chr.22: 29331783-29331783 on Build GRCh38
rs755652925 Chr.22: 29331747-29331750 on Build GRCh38
rs746583431 Chr.22: 29331885-29331885 on Build GRCh38
rs745363903 Chr.22: 29331950-29331950 on Build GRCh38
rs767707068 Chr.22: 29331830-29331830 on Build GRCh38
rs373344843 Chr.22: 29331932-29331932 on Build GRCh38
rs780608770 Chr.22: 29331949-29331949 on Build GRCh38
rs748511525 Chr.22: 29331878-29331878 on Build GRCh38
rs996045088 Chr.22: 29331787-29331787 on Build GRCh38
rs757462238 Chr.22: 29331829-29331829 on Build GRCh38
rs751840443 Chr.22: 29331821-29331821 on Build GRCh38
rs34740217 Chr.22: 29331851-29331852 on Build GRCh38
rs1019002619 Chr.22: 29331759-29331759 on Build GRCh38
rs756186368 Chr.22: 29331841-29331841 on Build GRCh38
rs764892604 Chr.22: 29331801-29331801 on Build GRCh38
rs767796976 Chr.22: 29331898-29331898 on Build GRCh38
rs200164791 Chr.22: 29331880-29331880 on Build GRCh38
rs776653588 Chr.22: 29331891-29331891 on Build GRCh38
rs779450787 Chr.22: 29331921-29331921 on Build GRCh38
rs770596350 Chr.22: 29331945-29331945 on Build GRCh38
rs200984005 Chr.22: 29331901-29331901 on Build GRCh38
rs372200882 Chr.22: 29331761-29331761 on Build GRCh38
rs866036616 Chr.22: 29331803-29331803 on Build GRCh38
rs777571819 Chr.22: 29331933-29331933 on Build GRCh38
rs766480290 Chr.22: 29331777-29331777 on Build GRCh38
rs750580486 Chr.22: 29331900-29331900 on Build GRCh38

View Full Product Details