Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 56719852-56720093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf168
Entrez Gene ID
199920
Gene Name
chromosome 1 open reading frame 168
Gene Aliases
-
UniGene
Hs.437655
Gene Chromosome Location
Chr.1: 56718804-56831985 on Build GRCh38
Transcripts
NM_001004303.4 XM_005270584.3 XM_011540898.1 XM_011540899.1 XM_011540904.2 XM_011540906.2 XM_017000544.1 XM_017000545.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4780479 c.2132-3T>A p.? Chr.1: 56720058-56720058 on Build GRCh38
3786250 c.2140G>A p.V714M Chr.1: 56720047-56720047 on Build GRCh38
6056623 c.2158G>C p.D720H Chr.1: 56720029-56720029 on Build GRCh38
3966653 c.2148T>C p.I716I Chr.1: 56720039-56720039 on Build GRCh38
3751317 c.2145C>A p.L715L Chr.1: 56720042-56720042 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758353226 Chr.1: 56720042-56720042 on Build GRCh38
rs570842875 Chr.1: 56720034-56720034 on Build GRCh38
rs776019686 Chr.1: 56719978-56719978 on Build GRCh38
rs369781349 Chr.1: 56719971-56719971 on Build GRCh38
rs112183545 Chr.1: 56720044-56720044 on Build GRCh38
rs757524202 Chr.1: 56720046-56720046 on Build GRCh38
rs765904420 Chr.1: 56720001-56720001 on Build GRCh38
rs537597313 Chr.1: 56720017-56720017 on Build GRCh38
rs200605296 Chr.1: 56720048-56720048 on Build GRCh38
rs566601372 Chr.1: 56719895-56719895 on Build GRCh38
rs769071318 Chr.1: 56719988-56719988 on Build GRCh38
rs774517636 Chr.1: 56719989-56719989 on Build GRCh38
rs199667769 Chr.1: 56720053-56720053 on Build GRCh38
rs961252605 Chr.1: 56719949-56719949 on Build GRCh38
rs1028193946 Chr.1: 56719893-56719893 on Build GRCh38
rs959235336 Chr.1: 56719934-56719934 on Build GRCh38
rs546605176 Chr.1: 56719886-56719886 on Build GRCh38
rs149466635 Chr.1: 56720021-56720021 on Build GRCh38
rs780097934 Chr.1: 56720060-56720060 on Build GRCh38
rs962225497 Chr.1: 56719914-56719914 on Build GRCh38
rs982023304 Chr.1: 56719918-56719918 on Build GRCh38
rs538759371 Chr.1: 56719900-56719900 on Build GRCh38
rs746074432 Chr.1: 56720051-56720051 on Build GRCh38
rs1015813406 Chr.1: 56719928-56719928 on Build GRCh38
rs760376294 Chr.1: 56719995-56719995 on Build GRCh38
rs558650643 Chr.1: 56719908-56719908 on Build GRCh38
rs753407888 Chr.1: 56720002-56720002 on Build GRCh38
rs763450369 Chr.1: 56719980-56719980 on Build GRCh38
rs752878642 Chr.1: 56719889-56719889 on Build GRCh38
rs575405647 Chr.1: 56719935-56719935 on Build GRCh38

View Full Product Details