Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 135285524-135285785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHI1
Entrez Gene ID
54806
Gene Name
Abelson helper integration site 1
Gene Aliases
AHI-1, JBTS3, ORF1, dJ71N10.1
UniGene
Hs.386684
Gene Chromosome Location
Chr.6: 135283972-135497775 on Build GRCh38
Transcripts
NM_001134830.1 NM_001134831.1 NM_017651.4 XM_011535910.2 XM_011535911.2 XM_017010977.1 XM_017010981.1 XM_017010982.1

SNP Detail

SNP ID Chromosome Location
rs1008098221 Chr.6: 135285638-135285638 on Build GRCh38
rs376845101 Chr.6: 135285609-135285609 on Build GRCh38
rs777982682 Chr.6: 135285680-135285680 on Build GRCh38
rs546334430 Chr.6: 135285673-135285674 on Build GRCh38
rs886061108 Chr.6: 135285577-135285577 on Build GRCh38
rs751910136 Chr.6: 135285640-135285640 on Build GRCh38
rs761218732 Chr.6: 135285623-135285623 on Build GRCh38
rs11970282 Chr.6: 135285753-135285753 on Build GRCh38
rs766922082 Chr.6: 135285639-135285639 on Build GRCh38
rs375597238 Chr.6: 135285567-135285567 on Build GRCh38
rs536580823 Chr.6: 135285565-135285565 on Build GRCh38
rs750018759 Chr.6: 135285721-135285721 on Build GRCh38
rs377458159 Chr.6: 135285666-135285666 on Build GRCh38
rs1027715985 Chr.6: 135285728-135285728 on Build GRCh38
rs775032489 Chr.6: 135285611-135285611 on Build GRCh38
rs201034412 Chr.6: 135285663-135285663 on Build GRCh38
rs755318468 Chr.6: 135285644-135285644 on Build GRCh38
rs545604059 Chr.6: 135285729-135285729 on Build GRCh38
rs756467270 Chr.6: 135285671-135285671 on Build GRCh38
rs763223260 Chr.6: 135285651-135285652 on Build GRCh38
rs764872326 Chr.6: 135285626-135285626 on Build GRCh38
rs1054190332 Chr.6: 135285577-135285577 on Build GRCh38
rs864622110 Chr.6: 135285652-135285652 on Build GRCh38
rs752910557 Chr.6: 135285636-135285640 on Build GRCh38
rs781417999 Chr.6: 135285650-135285650 on Build GRCh38
rs996603518 Chr.6: 135285714-135285714 on Build GRCh38
rs371523700 Chr.6: 135285604-135285604 on Build GRCh38
rs9494209 Chr.6: 135285617-135285617 on Build GRCh38
rs763438365 Chr.6: 135285632-135285632 on Build GRCh38
rs892893239 Chr.6: 135285602-135285602 on Build GRCh38
rs148731488 Chr.6: 135285750-135285750 on Build GRCh38
rs1038679542 Chr.6: 135285699-135285699 on Build GRCh38
rs377643105 Chr.6: 135285636-135285636 on Build GRCh38
rs900186828 Chr.6: 135285725-135285725 on Build GRCh38
rs1030238107 Chr.6: 135285737-135285737 on Build GRCh38

View Full Product Details