Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151233809-151234078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHPF2
Entrez Gene ID
54480
Gene Name
chondroitin polymerizing factor 2
Gene Aliases
CSGLCA-T, CSGlcAT, ChSy-3, chPF-2
UniGene
Hs.647084
Gene Chromosome Location
Chr.7: 151232489-151238827 on Build GRCh38
Transcripts
NM_001284295.1 NM_019015.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6321285 c.57G>T p.G19G Chr.7: 151234068-151234068 on Build GRCh38
5062306 c.3G>A p.M1I Chr.7: 151234014-151234014 on Build GRCh38
3261943 c.5G>A p.R2Q Chr.7: 151234016-151234016 on Build GRCh38
1241009 c.38C>T p.A13V Chr.7: 151234049-151234049 on Build GRCh38
3698034 c.27G>A p.L9L Chr.7: 151234038-151234038 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs559437863 Chr.7: 151233857-151233857 on Build GRCh38
rs146605020 Chr.7: 151234041-151234041 on Build GRCh38
rs748670587 Chr.7: 151234015-151234015 on Build GRCh38
rs772517432 Chr.7: 151234016-151234016 on Build GRCh38
rs779254993 Chr.7: 151234006-151234006 on Build GRCh38
rs761057741 Chr.7: 151234023-151234023 on Build GRCh38
rs751420591 Chr.7: 151233962-151233962 on Build GRCh38
rs928151717 Chr.7: 151234057-151234057 on Build GRCh38
rs1021961619 Chr.7: 151233960-151233960 on Build GRCh38
rs184568642 Chr.7: 151233851-151233851 on Build GRCh38
rs768742623 Chr.7: 151233989-151233989 on Build GRCh38
rs771442256 Chr.7: 151234033-151234033 on Build GRCh38
rs187857941 Chr.7: 151233883-151233883 on Build GRCh38
rs956370440 Chr.7: 151233914-151233914 on Build GRCh38
rs984490608 Chr.7: 151233939-151233939 on Build GRCh38
rs941134046 Chr.7: 151233872-151233872 on Build GRCh38
rs1029577795 Chr.7: 151234028-151234028 on Build GRCh38
rs768979863 Chr.7: 151233902-151233902 on Build GRCh38
rs528478295 Chr.7: 151233867-151233867 on Build GRCh38
rs368750482 Chr.7: 151234042-151234042 on Build GRCh38
rs981027305 Chr.7: 151234008-151234008 on Build GRCh38
rs763868595 Chr.7: 151234046-151234046 on Build GRCh38
rs2252694 Chr.7: 151233879-151233879 on Build GRCh38
rs185091725 Chr.7: 151233979-151233979 on Build GRCh38
rs776380320 Chr.7: 151234039-151234039 on Build GRCh38
rs192985599 Chr.7: 151233933-151233933 on Build GRCh38
rs778712444 Chr.7: 151233908-151233908 on Build GRCh38
rs890275387 Chr.7: 151233828-151233828 on Build GRCh38
rs749536446 Chr.7: 151233981-151233981 on Build GRCh38
rs570512685 Chr.7: 151234043-151234043 on Build GRCh38
rs146474219 Chr.7: 151233850-151233850 on Build GRCh38

View Full Product Details