Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 26916843-26917100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP4-AS1
Entrez Gene ID
147429
Gene Name
AQP4 antisense RNA 1
Gene Aliases
C18orf16, CHST9-AS1
UniGene
Hs.559280
Gene Chromosome Location
Chr.18: 26865308-26935946 on Build GRCh38
Transcripts
NR_026908.1 NM_001256316.1 NM_031422.5 XM_005258362.3 XM_017026033.1 XM_017026034.1

Gene Symbol
CHST9
Entrez Gene ID
83539
Gene Name
carbohydrate sulfotransferase 9
Gene Aliases
GALNAC4ST-2, GalNAc4ST2
UniGene
Hs.44584
Gene Chromosome Location
Chr.18: 26908754-27185364 on Build GRCh38
Transcripts
NR_026908.1 NM_001256316.1 NM_031422.5 XM_005258362.3 XM_017026033.1 XM_017026034.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1201182 c.500A>C p.K167T Chr.18: 26917091-26917091 on Build GRCh38
438005 c.413G>C p.G138A Chr.18: 26916923-26916923 on Build GRCh38
5651173 c.726C>T p.Y242Y Chr.18: 26916865-26916865 on Build GRCh38
438006 c.497A>C p.N166T Chr.18: 26917094-26917094 on Build GRCh38
1201178 c.601C>A p.H201N Chr.18: 26916990-26916990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747242394 Chr.18: 26916914-26916914 on Build GRCh38
rs773195205 Chr.18: 26916988-26916988 on Build GRCh38
rs771001978 Chr.18: 26916916-26916916 on Build GRCh38
rs418546 Chr.18: 26916953-26916953 on Build GRCh38
rs1044303789 Chr.18: 26916971-26916971 on Build GRCh38
rs748675373 Chr.18: 26916867-26916867 on Build GRCh38
rs745900618 Chr.18: 26916927-26916927 on Build GRCh38
rs765879879 Chr.18: 26917013-26917013 on Build GRCh38
rs868146176 Chr.18: 26916925-26916925 on Build GRCh38
rs769018799 Chr.18: 26917045-26917049 on Build GRCh38
rs1056860735 Chr.18: 26917007-26917007 on Build GRCh38
rs572655578 Chr.18: 26916913-26916913 on Build GRCh38
rs753556095 Chr.18: 26917022-26917022 on Build GRCh38
rs760160843 Chr.18: 26916891-26916891 on Build GRCh38
rs770018817 Chr.18: 26916944-26916944 on Build GRCh38
rs537543732 Chr.18: 26917032-26917032 on Build GRCh38
rs758732536 Chr.18: 26916903-26916903 on Build GRCh38
rs779350300 Chr.18: 26916863-26916863 on Build GRCh38
rs764768308 Chr.18: 26917024-26917024 on Build GRCh38
rs761879588 Chr.18: 26916956-26916956 on Build GRCh38
rs551975385 Chr.18: 26916940-26916940 on Build GRCh38
rs138794390 Chr.18: 26916982-26916982 on Build GRCh38
rs778268195 Chr.18: 26917059-26917059 on Build GRCh38
rs751808114 Chr.18: 26917064-26917064 on Build GRCh38
rs758963483 Chr.18: 26917058-26917058 on Build GRCh38
rs758794791 Chr.18: 26917023-26917023 on Build GRCh38
rs760531540 Chr.18: 26917006-26917006 on Build GRCh38
rs776920325 Chr.18: 26916917-26916917 on Build GRCh38
rs182429803 Chr.18: 26916949-26916949 on Build GRCh38
rs59171427 Chr.18: 26916985-26916985 on Build GRCh38

View Full Product Details