Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 67853782-67854053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPTF
Entrez Gene ID
2186
Gene Name
bromodomain PHD finger transcription factor
Gene Aliases
FAC1, FALZ, NURF301
UniGene
Hs.444200
Gene Chromosome Location
Chr.17: 67825517-67984378 on Build GRCh38
Transcripts
NM_004459.6 NM_182641.3 XM_005257150.3 XM_005257151.3 XM_005257152.2 XM_005257153.3 XM_005257154.3 XM_005257155.3 XM_005257156.3 XM_005257157.3 XM_005257158.3 XM_005257159.2 XM_005257160.2 XM_005257161.3 XM_011524520.2 XM_011524521.2 XM_011524522.2 XM_011524523.2 XM_011524525.2 XM_011524526.2 XM_017024353.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4665988 c.671T>C p.I224T Chr.17: 67853997-67853997 on Build GRCh38
4889506 c.660A>G p.E220E Chr.17: 67853986-67853986 on Build GRCh38
4665989 c.671T>C p.I224T Chr.17: 67853997-67853997 on Build GRCh38
4889507 c.660A>G p.E220E Chr.17: 67853986-67853986 on Build GRCh38
1189546 c.644G>A p.R215H Chr.17: 67853970-67853970 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562356766 Chr.17: 67853926-67853926 on Build GRCh38
rs752678842 Chr.17: 67853993-67853993 on Build GRCh38
rs1006936874 Chr.17: 67853862-67853862 on Build GRCh38
rs927797195 Chr.17: 67854001-67854001 on Build GRCh38
rs371591228 Chr.17: 67853896-67853896 on Build GRCh38
rs775833597 Chr.17: 67853925-67853925 on Build GRCh38
rs750466723 Chr.17: 67853967-67853967 on Build GRCh38
rs544473191 Chr.17: 67853914-67853914 on Build GRCh38
rs1042445958 Chr.17: 67853829-67853829 on Build GRCh38
rs762064359 Chr.17: 67853955-67853955 on Build GRCh38
rs372870037 Chr.17: 67854022-67854022 on Build GRCh38
rs528331492 Chr.17: 67853875-67853875 on Build GRCh38
rs981677727 Chr.17: 67853888-67853888 on Build GRCh38
rs754887842 Chr.17: 67853970-67853970 on Build GRCh38
rs781118428 Chr.17: 67853981-67853981 on Build GRCh38
rs564005359 Chr.17: 67853864-67853864 on Build GRCh38
rs902800448 Chr.17: 67853835-67853835 on Build GRCh38
rs369642020 Chr.17: 67853921-67853921 on Build GRCh38
rs972791629 Chr.17: 67853904-67853904 on Build GRCh38
rs867965880 Chr.17: 67853881-67853881 on Build GRCh38
rs190692658 Chr.17: 67854000-67854000 on Build GRCh38
rs998831795 Chr.17: 67853837-67853837 on Build GRCh38
rs530391038 Chr.17: 67853831-67853831 on Build GRCh38
rs368796044 Chr.17: 67853918-67853918 on Build GRCh38
rs74842951 Chr.17: 67853913-67853913 on Build GRCh38
rs764425294 Chr.17: 67853934-67853934 on Build GRCh38
rs748939899 Chr.17: 67853916-67853916 on Build GRCh38
rs773241506 Chr.17: 67853896-67853900 on Build GRCh38
rs754108327 Chr.17: 67853951-67853951 on Build GRCh38
rs770400803 Chr.17: 67853917-67853917 on Build GRCh38
rs946421964 Chr.17: 67853969-67853969 on Build GRCh38
rs914763670 Chr.17: 67853937-67853937 on Build GRCh38
rs2046323 Chr.17: 67853811-67853811 on Build GRCh38
rs186986885 Chr.17: 67853810-67853810 on Build GRCh38
rs777692636 Chr.17: 67854019-67854019 on Build GRCh38
rs954181866 Chr.17: 67853861-67853861 on Build GRCh38
rs552272446 Chr.17: 67853849-67853849 on Build GRCh38
rs765262835 Chr.17: 67853964-67853964 on Build GRCh38
rs772569850 Chr.17: 67853923-67853923 on Build GRCh38
rs762989222 Chr.17: 67853973-67853975 on Build GRCh38
rs754695790 Chr.17: 67853902-67853902 on Build GRCh38
rs769576037 Chr.17: 67853907-67853907 on Build GRCh38

View Full Product Details