Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 134506940-134507207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP63
Entrez Gene ID
80254
Gene Name
centrosomal protein 63
Gene Aliases
SCKL6
UniGene
Hs.443301
Gene Chromosome Location
Chr.3: 134485733-134629607 on Build GRCh38
Transcripts
NM_001042383.1 NM_001042384.1 NM_001042400.1 NM_025180.3 XM_005247795.4 XM_005247797.3 XM_005247799.3 XM_006713760.3 XM_011513194.2 XM_017007244.1 XM_017007245.1 XM_017007246.1 XM_017007247.1 XM_017007248.1 XM_017007249.1 XM_017007250.1 XM_017007251.1 XM_017007252.1 XM_017007253.1 XM_017007254.1 XM_017007255.1 XM_017007256.1 XM_017007257.1 XM_017007258.1 XM_017007259.1 XM_017007260.1 XM_017007261.1 XM_017007262.1 XM_017007263.1 XM_017007264.1 XM_017007265.1 XM_017007266.1 XM_017007267.1 XM_017007268.1 XM_017007269.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
353174 c.54G>C p.L18F Chr.3: 134507118-134507118 on Build GRCh38
6188997 c.97G>T p.D33Y Chr.3: 134507161-134507161 on Build GRCh38
6011213 c.87G>T p.M29I Chr.3: 134507151-134507151 on Build GRCh38
3759777 c.63T>C p.C21C Chr.3: 134507127-134507127 on Build GRCh38
6627118 c.136C>T p.R46C Chr.3: 134507200-134507200 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371227658 Chr.3: 134507072-134507072 on Build GRCh38
rs535018578 Chr.3: 134506987-134506987 on Build GRCh38
rs754676267 Chr.3: 134507149-134507149 on Build GRCh38
rs756434510 Chr.3: 134507079-134507079 on Build GRCh38
rs778851332 Chr.3: 134507186-134507187 on Build GRCh38
rs759695147 Chr.3: 134507112-134507112 on Build GRCh38
rs146294786 Chr.3: 134507186-134507186 on Build GRCh38
rs769757771 Chr.3: 134507066-134507066 on Build GRCh38
rs752514586 Chr.3: 134507164-134507164 on Build GRCh38
rs377250952 Chr.3: 134507148-134507148 on Build GRCh38
rs775917972 Chr.3: 134507121-134507121 on Build GRCh38
rs920083851 Chr.3: 134507062-134507062 on Build GRCh38
rs749449399 Chr.3: 134507161-134507161 on Build GRCh38
rs201362940 Chr.3: 134507154-134507154 on Build GRCh38
rs116793561 Chr.3: 134507063-134507063 on Build GRCh38
rs762439392 Chr.3: 134507140-134507140 on Build GRCh38
rs768963628 Chr.3: 134507174-134507174 on Build GRCh38
rs112759722 Chr.3: 134507135-134507135 on Build GRCh38
rs757413340 Chr.3: 134507097-134507098 on Build GRCh38
rs142072867 Chr.3: 134507165-134507165 on Build GRCh38
rs774435268 Chr.3: 134507099-134507099 on Build GRCh38
rs9827878 Chr.3: 134507127-134507127 on Build GRCh38
rs766439572 Chr.3: 134507033-134507033 on Build GRCh38
rs901780159 Chr.3: 134507015-134507015 on Build GRCh38
rs575090612 Chr.3: 134507073-134507073 on Build GRCh38
rs200369496 Chr.3: 134507125-134507125 on Build GRCh38
rs997169920 Chr.3: 134507056-134507056 on Build GRCh38
rs577557093 Chr.3: 134507145-134507145 on Build GRCh38
rs770960496 Chr.3: 134507096-134507096 on Build GRCh38
rs777655393 Chr.3: 134507167-134507167 on Build GRCh38
rs753982244 Chr.3: 134507096-134507098 on Build GRCh38
rs764450188 Chr.3: 134507079-134507080 on Build GRCh38
rs111472578 Chr.3: 134507043-134507043 on Build GRCh38
rs747964338 Chr.3: 134507065-134507065 on Build GRCh38
rs1038583024 Chr.3: 134506988-134506988 on Build GRCh38
rs772285151 Chr.3: 134507120-134507120 on Build GRCh38
rs572203890 Chr.3: 134506972-134506972 on Build GRCh38
rs372731094 Chr.3: 134507128-134507128 on Build GRCh38

View Full Product Details