Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 69599595-69599830 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AUTS2
Entrez Gene ID
26053
Gene Name
autism susceptibility candidate 2
Gene Aliases
FBRSL2, MRD26
UniGene
Hs.21631
Gene Chromosome Location
Chr.7: 69598475-70793068 on Build GRCh38
Transcripts
NM_001127231.2 NM_001127232.2 NM_015570.3 XM_011516010.2 XM_011516011.2 XM_011516012.2 XM_011516013.2 XM_011516014.2 XM_017011951.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4646292 c.113G>A p.G38D Chr.7: 69599766-69599766 on Build GRCh38
3750042 c.11C>T p.P4L Chr.7: 69599664-69599664 on Build GRCh38
6608927 c.166A>G p.K56E Chr.7: 69599819-69599819 on Build GRCh38
4635529 c.15G>A p.T5T Chr.7: 69599668-69599668 on Build GRCh38
4635528 c.15G>A p.T5T Chr.7: 69599668-69599668 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762676699 Chr.7: 69599755-69599778 on Build GRCh38
rs945733678 Chr.7: 69599781-69599781 on Build GRCh38
rs911513762 Chr.7: 69599771-69599771 on Build GRCh38
rs777541048 Chr.7: 69599782-69599782 on Build GRCh38
rs752490672 Chr.7: 69599750-69599773 on Build GRCh38
rs1041469078 Chr.7: 69599787-69599787 on Build GRCh38
rs1041341061 Chr.7: 69599668-69599668 on Build GRCh38
rs891065709 Chr.7: 69599633-69599633 on Build GRCh38
rs756779190 Chr.7: 69599784-69599784 on Build GRCh38
rs746774405 Chr.7: 69599806-69599806 on Build GRCh38
rs780512031 Chr.7: 69599755-69599755 on Build GRCh38
rs775225727 Chr.7: 69599802-69599802 on Build GRCh38
rs200189077 Chr.7: 69599664-69599664 on Build GRCh38
rs910011165 Chr.7: 69599731-69599731 on Build GRCh38
rs3735260 Chr.7: 69599651-69599651 on Build GRCh38
rs771784577 Chr.7: 69599801-69599801 on Build GRCh38
rs1051354561 Chr.7: 69599761-69599761 on Build GRCh38
rs976017092 Chr.7: 69599743-69599743 on Build GRCh38
rs752423410 Chr.7: 69599765-69599765 on Build GRCh38
rs745753819 Chr.7: 69599798-69599798 on Build GRCh38
rs755792920 Chr.7: 69599780-69599780 on Build GRCh38
rs934214411 Chr.7: 69599759-69599759 on Build GRCh38
rs913230763 Chr.7: 69599727-69599727 on Build GRCh38
rs530867061 Chr.7: 69599740-69599740 on Build GRCh38
rs534062236 Chr.7: 69599612-69599612 on Build GRCh38
rs748798665 Chr.7: 69599783-69599783 on Build GRCh38
rs904301389 Chr.7: 69599794-69599794 on Build GRCh38
rs768911898 Chr.7: 69599810-69599810 on Build GRCh38
rs962834047 Chr.7: 69599733-69599733 on Build GRCh38
rs778785768 Chr.7: 69599797-69599797 on Build GRCh38

View Full Product Details