Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7193835-7194100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLG4
Entrez Gene ID
1742
Gene Name
discs large MAGUK scaffold protein 4
Gene Aliases
PSD95, SAP-90, SAP90
UniGene
Hs.463928
Gene Chromosome Location
Chr.17: 7189890-7220050 on Build GRCh38
Transcripts
NM_001128827.2 NM_001321074.1 NM_001321075.1 NM_001321076.1 NM_001321077.1 NM_001365.4 NR_135527.1 XM_005256491.1 XM_011523698.1 XM_011523699.2 XM_011523702.1 XM_017024288.1 XM_017024289.1 XM_017024290.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3521558 c.1496T>G p.L499* Chr.17: 7193974-7193974 on Build GRCh38
983776 c.1644+2T>C p.? Chr.17: 7193962-7193962 on Build GRCh38
3521559 c.1634T>G p.L545* Chr.17: 7193974-7193974 on Build GRCh38
4569239 c.1610T>C p.V537A Chr.17: 7193998-7193998 on Build GRCh38
4569238 c.1472T>C p.V491A Chr.17: 7193998-7193998 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765504973 Chr.17: 7194024-7194024 on Build GRCh38
rs750859837 Chr.17: 7193954-7193954 on Build GRCh38
rs200014209 Chr.17: 7193903-7193903 on Build GRCh38
rs774990966 Chr.17: 7193969-7193969 on Build GRCh38
rs757521916 Chr.17: 7193885-7193885 on Build GRCh38
rs569839177 Chr.17: 7193949-7193949 on Build GRCh38
rs758606292 Chr.17: 7193895-7193895 on Build GRCh38
rs776878914 Chr.17: 7193932-7193933 on Build GRCh38
rs202090862 Chr.17: 7193893-7193893 on Build GRCh38
rs547798290 Chr.17: 7193884-7193884 on Build GRCh38
rs780375296 Chr.17: 7194035-7194035 on Build GRCh38
rs751720862 Chr.17: 7194042-7194042 on Build GRCh38
rs372671064 Chr.17: 7194068-7194068 on Build GRCh38
rs78879704 Chr.17: 7194021-7194021 on Build GRCh38
rs1043270332 Chr.17: 7193894-7193894 on Build GRCh38
rs747029006 Chr.17: 7193906-7193906 on Build GRCh38
rs372076014 Chr.17: 7193953-7193953 on Build GRCh38
rs767003847 Chr.17: 7193854-7193854 on Build GRCh38
rs750735517 Chr.17: 7194025-7194025 on Build GRCh38
rs753267248 Chr.17: 7193869-7193869 on Build GRCh38
rs762285058 Chr.17: 7194023-7194023 on Build GRCh38
rs546567784 Chr.17: 7193922-7193922 on Build GRCh38
rs2586536 Chr.17: 7194047-7194047 on Build GRCh38
rs777813154 Chr.17: 7193932-7193932 on Build GRCh38
rs759109334 Chr.17: 7193950-7193950 on Build GRCh38
rs764611632 Chr.17: 7194013-7194013 on Build GRCh38
rs974617356 Chr.17: 7194008-7194008 on Build GRCh38
rs964570324 Chr.17: 7193956-7193956 on Build GRCh38
rs760126196 Chr.17: 7194012-7194012 on Build GRCh38
rs371743350 Chr.17: 7194028-7194028 on Build GRCh38
rs114042970 Chr.17: 7193879-7193879 on Build GRCh38
rs142609608 Chr.17: 7193934-7193934 on Build GRCh38
rs758708454 Chr.17: 7194026-7194026 on Build GRCh38
rs770613600 Chr.17: 7193943-7193943 on Build GRCh38
rs1002079787 Chr.17: 7193897-7193897 on Build GRCh38
rs116629356 Chr.17: 7193894-7193894 on Build GRCh38

View Full Product Details