Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 165679573-165679800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSRNP3
Entrez Gene ID
80034
Gene Name
cysteine and serine rich nuclear protein 3
Gene Aliases
FAM130A2, PPP1R73, TAIP-2, TAIP2
UniGene
Hs.470479
Gene Chromosome Location
Chr.2: 165469647-165689407 on Build GRCh38
Transcripts
NM_001172173.1 NM_024969.3 XM_005246865.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
461314 c.1582C>G p.Q528E Chr.2: 165679577-165679577 on Build GRCh38
4086265 c.1651A>G p.S551G Chr.2: 165679646-165679646 on Build GRCh38
6636941 c.1735G>A p.V579M Chr.2: 165679730-165679730 on Build GRCh38
4086266 c.1657A>G p.I553V Chr.2: 165679652-165679652 on Build GRCh38
5584946 c.1578C>G p.H526Q Chr.2: 165679573-165679573 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771003387 Chr.2: 165679625-165679625 on Build GRCh38
rs768454412 Chr.2: 165679604-165679604 on Build GRCh38
rs760888237 Chr.2: 165679760-165679760 on Build GRCh38
rs781126827 Chr.2: 165679605-165679605 on Build GRCh38
rs754739038 Chr.2: 165679705-165679705 on Build GRCh38
rs768999404 Chr.2: 165679762-165679762 on Build GRCh38
rs772143221 Chr.2: 165679738-165679738 on Build GRCh38
rs934488204 Chr.2: 165679599-165679599 on Build GRCh38
rs748019473 Chr.2: 165679607-165679607 on Build GRCh38
rs774479697 Chr.2: 165679643-165679643 on Build GRCh38
rs537604087 Chr.2: 165679660-165679660 on Build GRCh38
rs751171426 Chr.2: 165679702-165679702 on Build GRCh38
rs765827808 Chr.2: 165679684-165679684 on Build GRCh38
rs777664315 Chr.2: 165679729-165679729 on Build GRCh38
rs769779209 Chr.2: 165679728-165679728 on Build GRCh38
rs764495569 Chr.2: 165679665-165679665 on Build GRCh38
rs757892024 Chr.2: 165679690-165679690 on Build GRCh38
rs772224445 Chr.2: 165679656-165679656 on Build GRCh38
rs760970772 Chr.2: 165679662-165679662 on Build GRCh38
rs968346417 Chr.2: 165679644-165679644 on Build GRCh38
rs780897912 Chr.2: 165679725-165679725 on Build GRCh38
rs142921916 Chr.2: 165679689-165679689 on Build GRCh38
rs867290492 Chr.2: 165679738-165679738 on Build GRCh38
rs779602441 Chr.2: 165679698-165679698 on Build GRCh38
rs548274315 Chr.2: 165679651-165679651 on Build GRCh38
rs61743584 Chr.2: 165679727-165679727 on Build GRCh38
rs528077010 Chr.2: 165679641-165679641 on Build GRCh38
rs199543465 Chr.2: 165679766-165679766 on Build GRCh38
rs754404268 Chr.2: 165679681-165679681 on Build GRCh38
rs200637688 Chr.2: 165679771-165679771 on Build GRCh38
rs746817797 Chr.2: 165679602-165679602 on Build GRCh38
rs762480588 Chr.2: 165679683-165679683 on Build GRCh38
rs146975789 Chr.2: 165679740-165679740 on Build GRCh38
rs1021572194 Chr.2: 165679628-165679628 on Build GRCh38
rs761772341 Chr.2: 165679730-165679730 on Build GRCh38
rs759647047 Chr.2: 165679649-165679649 on Build GRCh38

View Full Product Details