Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 162755551-162755822 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDR2
Entrez Gene ID
4921
Gene Name
discoidin domain receptor tyrosine kinase 2
Gene Aliases
MIG20a, NTRKR3, TKT, TYRO10
UniGene
Hs.275757
Gene Chromosome Location
Chr.1: 162631213-162783524 on Build GRCh38
Transcripts
NM_001014796.1 NM_006182.2 XM_006711344.3 XM_011509586.2 XM_011509587.2 XM_011509588.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3733082 c.664G>A p.G222R Chr.1: 162755762-162755762 on Build GRCh38
5735716 c.592G>A p.A198T Chr.1: 162755690-162755690 on Build GRCh38
6984799 c.566-2A>G p.? Chr.1: 162755662-162755662 on Build GRCh38
7427810 c.639T>C p.N213N Chr.1: 162755737-162755737 on Build GRCh38
1335757 c.601C>T p.Q201* Chr.1: 162755699-162755699 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759279024 Chr.1: 162755757-162755757 on Build GRCh38
rs760282956 Chr.1: 162755784-162755784 on Build GRCh38
rs578130186 Chr.1: 162755578-162755578 on Build GRCh38
rs1008158835 Chr.1: 162755682-162755682 on Build GRCh38
rs768606333 Chr.1: 162755690-162755690 on Build GRCh38
rs767109179 Chr.1: 162755622-162755622 on Build GRCh38
rs371241468 Chr.1: 162755601-162755602 on Build GRCh38
rs762600142 Chr.1: 162755729-162755729 on Build GRCh38
rs765780549 Chr.1: 162755788-162755788 on Build GRCh38
rs190639601 Chr.1: 162755789-162755789 on Build GRCh38
rs751226708 Chr.1: 162755614-162755614 on Build GRCh38
rs186645922 Chr.1: 162755571-162755571 on Build GRCh38
rs767100531 Chr.1: 162755768-162755768 on Build GRCh38
rs372660330 Chr.1: 162755631-162755631 on Build GRCh38
rs758800987 Chr.1: 162755660-162755660 on Build GRCh38
rs747324281 Chr.1: 162755689-162755689 on Build GRCh38
rs750765542 Chr.1: 162755652-162755652 on Build GRCh38
rs939944412 Chr.1: 162755584-162755584 on Build GRCh38
rs770407572 Chr.1: 162755601-162755601 on Build GRCh38
rs758962105 Chr.1: 162755619-162755619 on Build GRCh38
rs766826948 Chr.1: 162755793-162755793 on Build GRCh38
rs779593506 Chr.1: 162755638-162755638 on Build GRCh38
rs897863439 Chr.1: 162755612-162755612 on Build GRCh38
rs770824472 Chr.1: 162755730-162755730 on Build GRCh38
rs537213171 Chr.1: 162755595-162755595 on Build GRCh38
rs769683391 Chr.1: 162755713-162755713 on Build GRCh38
rs557254647 Chr.1: 162755708-162755708 on Build GRCh38
rs773858562 Chr.1: 162755732-162755732 on Build GRCh38
rs979423124 Chr.1: 162755599-162755600 on Build GRCh38
rs773746096 Chr.1: 162755697-162755697 on Build GRCh38
rs773159664 Chr.1: 162755717-162755717 on Build GRCh38
rs757878975 Chr.1: 162755634-162755634 on Build GRCh38
rs764925901 Chr.1: 162755778-162755778 on Build GRCh38
rs889257496 Chr.1: 162755676-162755676 on Build GRCh38
rs781298141 Chr.1: 162755691-162755691 on Build GRCh38
rs780075289 Chr.1: 162755685-162755685 on Build GRCh38
rs867004006 Chr.1: 162755693-162755693 on Build GRCh38

View Full Product Details