Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96267809-96268034 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIAO1
Entrez Gene ID
9391
Gene Name
cytosolic iron-sulfur assembly component 1
Gene Aliases
CIA1, WDR39
UniGene
Hs.12109
Gene Chromosome Location
Chr.2: 96266146-96274179 on Build GRCh38
Transcripts
NM_004804.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1023858 c.436C>T p.L146F Chr.2: 96267871-96267871 on Build GRCh38
6509572 c.457G>A p.V153I Chr.2: 96267892-96267892 on Build GRCh38
4996268 c.479C>T p.P160L Chr.2: 96267914-96267914 on Build GRCh38
5834512 c.483_484insG p.Q162fs*10 Chr.2: 96267918-96267919 on Build GRCh38
5834513 c.486_487insCC p.E163fs*4 Chr.2: 96267921-96267922 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201951574 Chr.2: 96267922-96267922 on Build GRCh38
rs900511645 Chr.2: 96267836-96267836 on Build GRCh38
rs756574386 Chr.2: 96267862-96267862 on Build GRCh38
rs766519074 Chr.2: 96267844-96267844 on Build GRCh38
rs779741693 Chr.2: 96267909-96267909 on Build GRCh38
rs769359653 Chr.2: 96267890-96267890 on Build GRCh38
rs759679157 Chr.2: 96267943-96267943 on Build GRCh38
rs777348944 Chr.2: 96267941-96267941 on Build GRCh38
rs751470231 Chr.2: 96267871-96267871 on Build GRCh38
rs374399730 Chr.2: 96267956-96267956 on Build GRCh38
rs759766043 Chr.2: 96267847-96267847 on Build GRCh38
rs953761555 Chr.2: 96267879-96267879 on Build GRCh38
rs373437838 Chr.2: 96267920-96267920 on Build GRCh38
rs537126505 Chr.2: 96267869-96267869 on Build GRCh38
rs931141471 Chr.2: 96267983-96267983 on Build GRCh38
rs1020430745 Chr.2: 96267835-96267835 on Build GRCh38
rs918460001 Chr.2: 96267965-96267965 on Build GRCh38
rs757205762 Chr.2: 96267872-96267872 on Build GRCh38
rs200422771 Chr.2: 96267957-96267957 on Build GRCh38
rs201103604 Chr.2: 96267936-96267936 on Build GRCh38
rs1003332443 Chr.2: 96267995-96267995 on Build GRCh38
rs189008080 Chr.2: 96267953-96267953 on Build GRCh38
rs771569260 Chr.2: 96267939-96267939 on Build GRCh38
rs765496479 Chr.2: 96267848-96267848 on Build GRCh38
rs868125888 Chr.2: 96267845-96267846 on Build GRCh38
rs927350496 Chr.2: 96268000-96268000 on Build GRCh38
rs752922921 Chr.2: 96267850-96267850 on Build GRCh38
rs985507273 Chr.2: 96267898-96267898 on Build GRCh38
rs765285433 Chr.2: 96267952-96267952 on Build GRCh38
rs966525448 Chr.2: 96267979-96267979 on Build GRCh38
rs200095914 Chr.2: 96267855-96267855 on Build GRCh38
rs377428847 Chr.2: 96267875-96267875 on Build GRCh38
rs528385159 Chr.2: 96267933-96267933 on Build GRCh38
rs370038210 Chr.2: 96267846-96267846 on Build GRCh38
rs373065541 Chr.2: 96267868-96267868 on Build GRCh38
rs139726148 Chr.2: 96267882-96267882 on Build GRCh38

View Full Product Details