Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 75564383-75564608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf26
Entrez Gene ID
152816
Gene Name
chromosome 4 open reading frame 26
Gene Aliases
AI2A4
UniGene
Hs.24510
Gene Chromosome Location
Chr.4: 75554997-75565893 on Build GRCh38
Transcripts
NM_001206981.1 NM_001257072.1 NM_178497.3 NR_046429.1 NR_046430.1 XM_011531668.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3605848 c.392C>T p.S131F Chr.4: 75564394-75564394 on Build GRCh38
5890000 c.433G>A p.A145T Chr.4: 75564435-75564435 on Build GRCh38
3380983 c.467C>A p.A156E Chr.4: 75564469-75564469 on Build GRCh38
2831658 c.468delA p.S159fs*>18 Chr.4: 75564470-75564470 on Build GRCh38
2831655 c.409A>G p.R137G Chr.4: 75564411-75564411 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200717133 Chr.4: 75564481-75564481 on Build GRCh38
rs771621604 Chr.4: 75564537-75564537 on Build GRCh38
rs771966707 Chr.4: 75564440-75564440 on Build GRCh38
rs770731754 Chr.4: 75564455-75564455 on Build GRCh38
rs776461175 Chr.4: 75564569-75564569 on Build GRCh38
rs756386294 Chr.4: 75564497-75564497 on Build GRCh38
rs192831647 Chr.4: 75564525-75564525 on Build GRCh38
rs751699183 Chr.4: 75564475-75564475 on Build GRCh38
rs138123570 Chr.4: 75564423-75564423 on Build GRCh38
rs778561995 Chr.4: 75564529-75564529 on Build GRCh38
rs766567294 Chr.4: 75564501-75564502 on Build GRCh38
rs377354901 Chr.4: 75564421-75564421 on Build GRCh38
rs764141250 Chr.4: 75564470-75564470 on Build GRCh38
rs763152187 Chr.4: 75564570-75564570 on Build GRCh38
rs748978797 Chr.4: 75564420-75564420 on Build GRCh38
rs759373305 Chr.4: 75564553-75564556 on Build GRCh38
rs751319059 Chr.4: 75564544-75564544 on Build GRCh38
rs773385942 Chr.4: 75564470-75564470 on Build GRCh38
rs763205114 Chr.4: 75564464-75564464 on Build GRCh38
rs752890853 Chr.4: 75564526-75564526 on Build GRCh38
rs750954612 Chr.4: 75564494-75564494 on Build GRCh38
rs773202196 Chr.4: 75564445-75564445 on Build GRCh38
rs961670750 Chr.4: 75564443-75564443 on Build GRCh38
rs746995232 Chr.4: 75564558-75564558 on Build GRCh38
rs747541499 Chr.4: 75564530-75564530 on Build GRCh38
rs188735683 Chr.4: 75564512-75564512 on Build GRCh38
rs777519994 Chr.4: 75564556-75564556 on Build GRCh38
rs116697757 Chr.4: 75564568-75564568 on Build GRCh38
rs748102550 Chr.4: 75564425-75564425 on Build GRCh38
rs775653502 Chr.4: 75564459-75564459 on Build GRCh38
rs767845966 Chr.4: 75564488-75564488 on Build GRCh38
rs373107558 Chr.4: 75564412-75564412 on Build GRCh38
rs763292297 Chr.4: 75564469-75564470 on Build GRCh38
rs964917900 Chr.4: 75564489-75564489 on Build GRCh38
rs760326382 Chr.4: 75564446-75564446 on Build GRCh38

View Full Product Details