Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1341133-1341380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAIAP3
Entrez Gene ID
8938
Gene Name
BAI1 associated protein 3
Gene Aliases
BAP3
UniGene
Hs.458427
Gene Chromosome Location
Chr.16: 1333605-1349441 on Build GRCh38
Transcripts
NM_001199096.1 NM_001199097.1 NM_001199098.1 NM_001199099.1 NM_001286464.1 NM_003933.4 XM_011522728.1 XM_011522729.1 XM_011522730.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4557085 c.714G>A p.K238K Chr.16: 1341367-1341367 on Build GRCh38
3377775 c.694C>T p.R232W Chr.16: 1341347-1341347 on Build GRCh38
6078379 c.603C>T p.V201V Chr.16: 1341158-1341158 on Build GRCh38
6369452 c.606C>A p.C202* Chr.16: 1341313-1341313 on Build GRCh38
318887 c.722G>T p.R241L Chr.16: 1341375-1341375 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769931870 Chr.16: 1341279-1341279 on Build GRCh38
rs767405132 Chr.16: 1341286-1341286 on Build GRCh38
rs141276912 Chr.16: 1341184-1341184 on Build GRCh38
rs747416397 Chr.16: 1341340-1341340 on Build GRCh38
rs373347609 Chr.16: 1341323-1341323 on Build GRCh38
rs202213122 Chr.16: 1341260-1341260 on Build GRCh38
rs761291680 Chr.16: 1341222-1341222 on Build GRCh38
rs142578030 Chr.16: 1341356-1341356 on Build GRCh38
rs137994295 Chr.16: 1341349-1341349 on Build GRCh38
rs752977035 Chr.16: 1341228-1341228 on Build GRCh38
rs749009798 Chr.16: 1341255-1341255 on Build GRCh38
rs762120629 Chr.16: 1341360-1341360 on Build GRCh38
rs777067992 Chr.16: 1341272-1341272 on Build GRCh38
rs748876413 Chr.16: 1341207-1341207 on Build GRCh38
rs978127870 Chr.16: 1341301-1341301 on Build GRCh38
rs945283697 Chr.16: 1341307-1341307 on Build GRCh38
rs746418572 Chr.16: 1341346-1341346 on Build GRCh38
rs758158159 Chr.16: 1341181-1341181 on Build GRCh38
rs779757203 Chr.16: 1341312-1341312 on Build GRCh38
rs116401495 Chr.16: 1341195-1341195 on Build GRCh38
rs115052205 Chr.16: 1341357-1341357 on Build GRCh38
rs779704420 Chr.16: 1341250-1341250 on Build GRCh38
rs750452604 Chr.16: 1341294-1341294 on Build GRCh38
rs537419858 Chr.16: 1341336-1341336 on Build GRCh38
rs1042340829 Chr.16: 1341314-1341314 on Build GRCh38
rs751402757 Chr.16: 1341284-1341284 on Build GRCh38
rs771345002 Chr.16: 1341341-1341341 on Build GRCh38
rs749966171 Chr.16: 1341246-1341246 on Build GRCh38
rs758526515 Chr.16: 1341201-1341202 on Build GRCh38
rs781537158 Chr.16: 1341345-1341345 on Build GRCh38
rs764173504 Chr.16: 1341283-1341283 on Build GRCh38
rs555666170 Chr.16: 1341270-1341270 on Build GRCh38
rs747295817 Chr.16: 1341363-1341365 on Build GRCh38
rs903167831 Chr.16: 1341325-1341325 on Build GRCh38
rs964311638 Chr.16: 1341243-1341243 on Build GRCh38
rs575914422 Chr.16: 1341337-1341337 on Build GRCh38
rs1012102124 Chr.16: 1341162-1341162 on Build GRCh38
rs768167147 Chr.16: 1341259-1341259 on Build GRCh38
rs755743825 Chr.16: 1341249-1341249 on Build GRCh38
rs761456098 Chr.16: 1341285-1341285 on Build GRCh38
rs763285037 Chr.16: 1341280-1341280 on Build GRCh38
rs375520815 Chr.16: 1341163-1341163 on Build GRCh38
rs145056617 Chr.16: 1341348-1341348 on Build GRCh38
rs757241660 Chr.16: 1341238-1341238 on Build GRCh38
rs371995818 Chr.16: 1341227-1341227 on Build GRCh38
rs34468534 Chr.16: 1341220-1341220 on Build GRCh38
rs200028638 Chr.16: 1341226-1341226 on Build GRCh38
rs751292454 Chr.16: 1341234-1341234 on Build GRCh38
rs760026043 Chr.16: 1341278-1341278 on Build GRCh38
rs779975248 Chr.16: 1341207-1341208 on Build GRCh38
rs769574111 Chr.16: 1341200-1341200 on Build GRCh38
rs775225174 Chr.16: 1341204-1341204 on Build GRCh38
rs893642792 Chr.16: 1341277-1341277 on Build GRCh38
rs997538561 Chr.16: 1341347-1341347 on Build GRCh38
rs747038087 Chr.16: 1341193-1341193 on Build GRCh38
rs200223867 Chr.16: 1341194-1341194 on Build GRCh38
rs745323518 Chr.16: 1341198-1341198 on Build GRCh38
rs376854970 Chr.16: 1341225-1341225 on Build GRCh38
rs576384264 Chr.16: 1341239-1341239 on Build GRCh38
rs763991844 Chr.16: 1341233-1341233 on Build GRCh38
rs557223354 Chr.16: 1341219-1341219 on Build GRCh38
rs762903308 Chr.16: 1341354-1341354 on Build GRCh38
rs553047589 Chr.16: 1341229-1341229 on Build GRCh38
rs75271587 Chr.16: 1341361-1341361 on Build GRCh38
rs756086952 Chr.16: 1341302-1341302 on Build GRCh38

View Full Product Details