Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 87662422-87662673 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMP1
Entrez Gene ID
1758
Gene Name
dentin matrix acidic phosphoprotein 1
Gene Aliases
ARHP, ARHR, DMP-1
UniGene
Hs.652366
Gene Chromosome Location
Chr.4: 87650302-87664361 on Build GRCh38
Transcripts
NM_001079911.2 NM_004407.3 XM_011531705.2 XM_011531706.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1753871 c.873C>G p.V291V Chr.4: 87662651-87662651 on Build GRCh38
3826410 c.868G>T p.E290* Chr.4: 87662646-87662646 on Build GRCh38
2957197 c.745G>T p.A249S Chr.4: 87662523-87662523 on Build GRCh38
3606856 c.795G>A p.R265R Chr.4: 87662573-87662573 on Build GRCh38
7182440 c.718A>G p.K240E Chr.4: 87662496-87662496 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773197665 Chr.4: 87662479-87662479 on Build GRCh38
rs751076227 Chr.4: 87662525-87662525 on Build GRCh38
rs190314299 Chr.4: 87662516-87662516 on Build GRCh38
rs201413886 Chr.4: 87662487-87662487 on Build GRCh38
rs35935935 Chr.4: 87662454-87662454 on Build GRCh38
rs867621609 Chr.4: 87662590-87662590 on Build GRCh38
rs760711288 Chr.4: 87662595-87662595 on Build GRCh38
rs969652844 Chr.4: 87662458-87662458 on Build GRCh38
rs766560937 Chr.4: 87662486-87662486 on Build GRCh38
rs759841451 Chr.4: 87662618-87662618 on Build GRCh38
rs756758638 Chr.4: 87662531-87662531 on Build GRCh38
rs924937956 Chr.4: 87662530-87662530 on Build GRCh38
rs565811736 Chr.4: 87662641-87662641 on Build GRCh38
rs752472758 Chr.4: 87662512-87662512 on Build GRCh38
rs373457215 Chr.4: 87662592-87662592 on Build GRCh38
rs765137056 Chr.4: 87662503-87662503 on Build GRCh38
rs771028926 Chr.4: 87662604-87662604 on Build GRCh38
rs748472892 Chr.4: 87662577-87662577 on Build GRCh38
rs571820459 Chr.4: 87662565-87662565 on Build GRCh38
rs867788427 Chr.4: 87662499-87662499 on Build GRCh38
rs139029727 Chr.4: 87662609-87662609 on Build GRCh38
rs141979823 Chr.4: 87662622-87662622 on Build GRCh38
rs751512932 Chr.4: 87662629-87662629 on Build GRCh38
rs888983238 Chr.4: 87662600-87662600 on Build GRCh38
rs765511837 Chr.4: 87662619-87662619 on Build GRCh38
rs377338877 Chr.4: 87662587-87662587 on Build GRCh38
rs760761426 Chr.4: 87662483-87662483 on Build GRCh38
rs1009092274 Chr.4: 87662535-87662535 on Build GRCh38
rs968385548 Chr.4: 87662597-87662597 on Build GRCh38
rs780778179 Chr.4: 87662553-87662553 on Build GRCh38
rs897755263 Chr.4: 87662631-87662631 on Build GRCh38
rs779258197 Chr.4: 87662570-87662570 on Build GRCh38
rs147552663 Chr.4: 87662502-87662502 on Build GRCh38
rs752558570 Chr.4: 87662620-87662620 on Build GRCh38
rs769382267 Chr.4: 87662568-87662568 on Build GRCh38
rs776792846 Chr.4: 87662614-87662614 on Build GRCh38
rs941952368 Chr.4: 87662617-87662617 on Build GRCh38
rs373051924 Chr.4: 87662452-87662452 on Build GRCh38
rs560963534 Chr.4: 87662561-87662561 on Build GRCh38
rs145237146 Chr.4: 87662593-87662593 on Build GRCh38

View Full Product Details