Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 154870668-154870893 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT8
Entrez Gene ID
9337
Gene Name
CCR4-NOT transcription complex subunit 8
Gene Aliases
CAF1, CALIF, Caf1b, POP2, hCAF1
UniGene
Hs.26703
Gene Chromosome Location
Chr.5: 154857952-154876792 on Build GRCh38
Transcripts
NM_001301073.1 NM_001301074.1 NM_001301075.1 NM_001301077.1 NM_001301080.1 NM_001301082.1 NM_001301083.1 NM_001301086.1 NM_004779.5 XM_005268527.2 XM_011537706.1 XM_011537708.2 XM_017010048.1 XM_017010049.1 XM_017010050.1 XM_017010051.1 XM_017010052.1 XM_017010053.1 XM_017010054.1 XM_017010055.1 XM_017010056.1 XM_017010057.1 XM_017010058.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3245716 c.403C>A p.H135N Chr.5: 154870752-154870752 on Build GRCh38
3853046 c.424A>T p.T142S Chr.5: 154870773-154870773 on Build GRCh38
1310883 c.319A>G p.M107V Chr.5: 154870668-154870668 on Build GRCh38
266272 c.353A>T p.N118I Chr.5: 154870702-154870702 on Build GRCh38
5026448 c.367T>C p.F123L Chr.5: 154870716-154870716 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776369775 Chr.5: 154870845-154870845 on Build GRCh38
rs143791180 Chr.5: 154870757-154870757 on Build GRCh38
rs761360552 Chr.5: 154870732-154870732 on Build GRCh38
rs533634377 Chr.5: 154870733-154870733 on Build GRCh38
rs760355696 Chr.5: 154870703-154870703 on Build GRCh38
rs929132272 Chr.5: 154870798-154870798 on Build GRCh38
rs759662862 Chr.5: 154870830-154870831 on Build GRCh38
rs773705720 Chr.5: 154870739-154870739 on Build GRCh38
rs773637490 Chr.5: 154870861-154870861 on Build GRCh38
rs375081243 Chr.5: 154870829-154870829 on Build GRCh38
rs769412049 Chr.5: 154870853-154870853 on Build GRCh38
rs368181033 Chr.5: 154870799-154870799 on Build GRCh38
rs763281414 Chr.5: 154870743-154870743 on Build GRCh38
rs778782009 Chr.5: 154870802-154870802 on Build GRCh38
rs998328732 Chr.5: 154870839-154870839 on Build GRCh38
rs747668873 Chr.5: 154870849-154870849 on Build GRCh38
rs374904720 Chr.5: 154870784-154870784 on Build GRCh38
rs768447949 Chr.5: 154870834-154870834 on Build GRCh38
rs371511333 Chr.5: 154870705-154870705 on Build GRCh38
rs776315780 Chr.5: 154870720-154870720 on Build GRCh38
rs371901945 Chr.5: 154870813-154870813 on Build GRCh38
rs752785048 Chr.5: 154870790-154870790 on Build GRCh38
rs751767336 Chr.5: 154870760-154870760 on Build GRCh38
rs755221649 Chr.5: 154870763-154870763 on Build GRCh38
rs746763311 Chr.5: 154870830-154870830 on Build GRCh38
rs758177720 Chr.5: 154870827-154870827 on Build GRCh38
rs768323343 Chr.5: 154870714-154870714 on Build GRCh38

View Full Product Details