Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 39164821-39165080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM32
Entrez Gene ID
203102
Gene Name
ADAM metallopeptidase domain 32
Gene Aliases
-
UniGene
Hs.521545
Gene Chromosome Location
Chr.8: 39107529-39284917 on Build GRCh38
Transcripts
NM_001313994.1 NM_145004.6 XM_006716298.2 XM_006716299.1 XM_011544433.1 XM_011544434.1 XM_017013183.1 XM_017013184.1 XM_017013185.1 XM_017013186.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6994493 c.665C>A p.S222* Chr.8: 39164834-39164834 on Build GRCh38
1099718 c.657T>C p.L219L Chr.8: 39164826-39164826 on Build GRCh38
5467438 c.687T>C p.V229V Chr.8: 39165050-39165050 on Build GRCh38
4927030 c.672C>T p.F224F Chr.8: 39165035-39165035 on Build GRCh38
5109932 c.666+3A>T p.? Chr.8: 39164838-39164838 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs912875731 Chr.8: 39165050-39165050 on Build GRCh38
rs748897987 Chr.8: 39164950-39164950 on Build GRCh38
rs754808674 Chr.8: 39165054-39165054 on Build GRCh38
rs890030209 Chr.8: 39164853-39164853 on Build GRCh38
rs751614865 Chr.8: 39165035-39165035 on Build GRCh38
rs753811738 Chr.8: 39165001-39165001 on Build GRCh38
rs374199610 Chr.8: 39164989-39164989 on Build GRCh38
rs904007383 Chr.8: 39165027-39165027 on Build GRCh38
rs774890524 Chr.8: 39164852-39164852 on Build GRCh38
rs186536952 Chr.8: 39164956-39164956 on Build GRCh38
rs10088854 Chr.8: 39165003-39165003 on Build GRCh38
rs766389395 Chr.8: 39165010-39165010 on Build GRCh38
rs909691144 Chr.8: 39164906-39164906 on Build GRCh38
rs957271769 Chr.8: 39165019-39165019 on Build GRCh38
rs557187963 Chr.8: 39164866-39164868 on Build GRCh38
rs553457255 Chr.8: 39165005-39165005 on Build GRCh38
rs982684052 Chr.8: 39164904-39164904 on Build GRCh38
rs905316916 Chr.8: 39164958-39164958 on Build GRCh38
rs777286867 Chr.8: 39164949-39164949 on Build GRCh38
rs539225710 Chr.8: 39164967-39164967 on Build GRCh38
rs1005837841 Chr.8: 39164903-39164903 on Build GRCh38
rs1022564649 Chr.8: 39165004-39165004 on Build GRCh38
rs1038353944 Chr.8: 39164924-39164924 on Build GRCh38
rs759887279 Chr.8: 39164857-39164857 on Build GRCh38
rs764407985 Chr.8: 39164991-39164991 on Build GRCh38
rs775932318 Chr.8: 39164981-39164981 on Build GRCh38
rs976480947 Chr.8: 39164858-39164859 on Build GRCh38
rs559202416 Chr.8: 39164973-39164973 on Build GRCh38
rs771381580 Chr.8: 39164846-39164846 on Build GRCh38
rs759757089 Chr.8: 39165023-39165023 on Build GRCh38
rs941227491 Chr.8: 39164928-39164928 on Build GRCh38
rs897180762 Chr.8: 39164932-39164932 on Build GRCh38
rs369075980 Chr.8: 39165024-39165024 on Build GRCh38
rs990038942 Chr.8: 39165025-39165025 on Build GRCh38
rs550225866 Chr.8: 39164915-39164915 on Build GRCh38
rs372226544 Chr.8: 39164859-39164859 on Build GRCh38
rs929873363 Chr.8: 39164896-39164896 on Build GRCh38

View Full Product Details