Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 72223856-72224117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC16
Entrez Gene ID
119504
Gene Name
anaphase promoting complex subunit 16
Gene Aliases
APC16, C10orf104, CENP-27, MSAG, bA570G20.3
UniGene
Hs.426296
Gene Chromosome Location
Chr.10: 72216000-72235860 on Build GRCh38
Transcripts
NM_001242546.1 NM_001242547.1 NM_001242548.1 NM_173473.3 NR_038391.1 NR_038392.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5792661 c.1A>G p.M1V Chr.10: 72223915-72223915 on Build GRCh38
1241982 c.18A>C p.S6S Chr.10: 72223932-72223932 on Build GRCh38
6272651 c.52G>T p.V18F Chr.10: 72223966-72223966 on Build GRCh38
5837268 c.114delC p.G41fs*6 Chr.10: 72224028-72224028 on Build GRCh38
4945686 c.142+5T>G p.? Chr.10: 72224061-72224061 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753474790 Chr.10: 72223966-72223966 on Build GRCh38
rs367571666 Chr.10: 72224083-72224083 on Build GRCh38
rs145495551 Chr.10: 72223974-72223974 on Build GRCh38
rs768147266 Chr.10: 72223936-72223936 on Build GRCh38
rs11553570 Chr.10: 72223940-72223940 on Build GRCh38
rs763764283 Chr.10: 72223955-72223955 on Build GRCh38
rs770005105 Chr.10: 72223904-72223904 on Build GRCh38
rs773458305 Chr.10: 72224016-72224016 on Build GRCh38
rs759996498 Chr.10: 72224046-72224046 on Build GRCh38
rs749533516 Chr.10: 72224025-72224025 on Build GRCh38
rs755602831 Chr.10: 72223943-72223943 on Build GRCh38
rs778744770 Chr.10: 72223999-72223999 on Build GRCh38
rs1047361551 Chr.10: 72224065-72224065 on Build GRCh38
rs745667154 Chr.10: 72224000-72224000 on Build GRCh38
rs561141916 Chr.10: 72224001-72224001 on Build GRCh38
rs771325361 Chr.10: 72224031-72224031 on Build GRCh38
rs776625808 Chr.10: 72223882-72223882 on Build GRCh38
rs368465315 Chr.10: 72224042-72224042 on Build GRCh38
rs748180945 Chr.10: 72224007-72224007 on Build GRCh38
rs763343055 Chr.10: 72223915-72223915 on Build GRCh38
rs779985832 Chr.10: 72224003-72224003 on Build GRCh38
rs147690332 Chr.10: 72224009-72224009 on Build GRCh38
rs374106007 Chr.10: 72223905-72223905 on Build GRCh38
rs147603657 Chr.10: 72223932-72223932 on Build GRCh38
rs776172325 Chr.10: 72224064-72224064 on Build GRCh38
rs772659116 Chr.10: 72224058-72224058 on Build GRCh38
rs748239814 Chr.10: 72223892-72223892 on Build GRCh38
rs201228064 Chr.10: 72223935-72223935 on Build GRCh38
rs764084573 Chr.10: 72224082-72224082 on Build GRCh38

View Full Product Details