Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7227958-7228187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DVL2
Entrez Gene ID
1856
Gene Name
dishevelled segment polarity protein 2
Gene Aliases
-
UniGene
Hs.118640
Gene Chromosome Location
Chr.17: 7225342-7234548 on Build GRCh38
Transcripts
NM_004422.2 XM_005256502.1 XM_017024298.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4580052 c.1092T>A p.T364T Chr.17: 7227987-7227987 on Build GRCh38
4909640 c.1100G>T p.R367L Chr.17: 7227979-7227979 on Build GRCh38
2976661 c.1075C>A p.P359T Chr.17: 7228004-7228004 on Build GRCh38
4069210 c.1076C>A p.P359H Chr.17: 7228003-7228003 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781418768 Chr.17: 7227979-7227979 on Build GRCh38
rs756414359 Chr.17: 7228084-7228084 on Build GRCh38
rs763785958 Chr.17: 7228050-7228050 on Build GRCh38
rs145282702 Chr.17: 7227980-7227980 on Build GRCh38
rs546945495 Chr.17: 7228135-7228135 on Build GRCh38
rs1026419544 Chr.17: 7228168-7228168 on Build GRCh38
rs1026789448 Chr.17: 7228109-7228109 on Build GRCh38
rs761710493 Chr.17: 7228053-7228053 on Build GRCh38
rs371100454 Chr.17: 7228101-7228101 on Build GRCh38
rs965064301 Chr.17: 7228162-7228162 on Build GRCh38
rs890310506 Chr.17: 7228069-7228069 on Build GRCh38
rs188458420 Chr.17: 7228157-7228157 on Build GRCh38
rs775018358 Chr.17: 7228026-7228026 on Build GRCh38
rs994558085 Chr.17: 7228161-7228161 on Build GRCh38
rs760443700 Chr.17: 7228040-7228040 on Build GRCh38
rs773689180 Chr.17: 7227994-7227994 on Build GRCh38
rs535994494 Chr.17: 7228064-7228064 on Build GRCh38
rs752853482 Chr.17: 7228077-7228077 on Build GRCh38
rs143084690 Chr.17: 7228004-7228004 on Build GRCh38
rs370287833 Chr.17: 7227987-7227987 on Build GRCh38
rs1053193592 Chr.17: 7228023-7228023 on Build GRCh38
rs866399492 Chr.17: 7228042-7228042 on Build GRCh38
rs751594569 Chr.17: 7228071-7228071 on Build GRCh38
rs767229371 Chr.17: 7228018-7228018 on Build GRCh38
rs533533057 Chr.17: 7228073-7228073 on Build GRCh38
rs62059164 Chr.17: 7228052-7228052 on Build GRCh38
rs1006291277 Chr.17: 7228144-7228144 on Build GRCh38
rs897029333 Chr.17: 7228099-7228099 on Build GRCh38
rs1044093221 Chr.17: 7228060-7228060 on Build GRCh38
rs137865252 Chr.17: 7227981-7227981 on Build GRCh38
rs777974146 Chr.17: 7228088-7228088 on Build GRCh38
rs905525697 Chr.17: 7228065-7228065 on Build GRCh38
rs781405704 Chr.17: 7228074-7228074 on Build GRCh38
rs766408450 Chr.17: 7228068-7228069 on Build GRCh38
rs1017342579 Chr.17: 7228141-7228141 on Build GRCh38
rs749743656 Chr.17: 7227997-7227997 on Build GRCh38
rs547857082 Chr.17: 7228014-7228014 on Build GRCh38
rs145138687 Chr.17: 7228013-7228013 on Build GRCh38
rs771612069 Chr.17: 7228001-7228001 on Build GRCh38
rs376020506 Chr.17: 7227982-7227982 on Build GRCh38
rs952135925 Chr.17: 7228128-7228128 on Build GRCh38

View Full Product Details