Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17137848-17138117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CECR5
Entrez Gene ID
27440
Gene Name
cat eye syndrome chromosome region, candidate 5
Gene Aliases
-
UniGene
Hs.26890
Gene Chromosome Location
Chr.22: 17137520-17165287 on Build GRCh38
Transcripts
NM_017829.5 NM_033070.2 XM_005261236.3 XM_011546126.2 XM_011546127.1 XM_017028781.1 XM_017028782.1 XM_017028783.1 XM_017028784.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6626025 c.1129G>A p.V377M Chr.22: 17138074-17138074 on Build GRCh38
1714123 c.1199_1200CC>TT p.A400V Chr.22: 17138093-17138094 on Build GRCh38
4461352 c.1112C>T p.S371F Chr.22: 17138091-17138091 on Build GRCh38
7081631 c.1251G>C p.K417N Chr.22: 17138042-17138042 on Build GRCh38
3964060 c.1267G>C p.E423Q Chr.22: 17138026-17138026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs939360781 Chr.22: 17137964-17137964 on Build GRCh38
rs751702424 Chr.22: 17137976-17137976 on Build GRCh38
rs549185431 Chr.22: 17138039-17138039 on Build GRCh38
rs930766135 Chr.22: 17137999-17137999 on Build GRCh38
rs757346994 Chr.22: 17138062-17138062 on Build GRCh38
rs372764350 Chr.22: 17138092-17138092 on Build GRCh38
rs748027190 Chr.22: 17137997-17137997 on Build GRCh38
rs199543754 Chr.22: 17138074-17138074 on Build GRCh38
rs972299076 Chr.22: 17137968-17137968 on Build GRCh38
rs778636242 Chr.22: 17138087-17138087 on Build GRCh38
rs927491854 Chr.22: 17137893-17137893 on Build GRCh38
rs746562473 Chr.22: 17138010-17138010 on Build GRCh38
rs564020230 Chr.22: 17137900-17137900 on Build GRCh38
rs772637918 Chr.22: 17138005-17138005 on Build GRCh38
rs764711946 Chr.22: 17138051-17138051 on Build GRCh38
rs747096920 Chr.22: 17138036-17138037 on Build GRCh38
rs746500177 Chr.22: 17138096-17138096 on Build GRCh38
rs35327402 Chr.22: 17138047-17138047 on Build GRCh38
rs770616219 Chr.22: 17138011-17138011 on Build GRCh38
rs377605731 Chr.22: 17138048-17138048 on Build GRCh38
rs142661724 Chr.22: 17137992-17137992 on Build GRCh38
rs777758471 Chr.22: 17138093-17138093 on Build GRCh38
rs980745103 Chr.22: 17137904-17137904 on Build GRCh38
rs377702374 Chr.22: 17137977-17137977 on Build GRCh38
rs759003653 Chr.22: 17137970-17137970 on Build GRCh38
rs927931882 Chr.22: 17137960-17137960 on Build GRCh38
rs749097009 Chr.22: 17137985-17137985 on Build GRCh38
rs772025344 Chr.22: 17138001-17138001 on Build GRCh38
rs759924679 Chr.22: 17138046-17138046 on Build GRCh38
rs369698082 Chr.22: 17137982-17137982 on Build GRCh38
rs751646054 Chr.22: 17138056-17138056 on Build GRCh38
rs139103323 Chr.22: 17138075-17138075 on Build GRCh38
rs764219571 Chr.22: 17137972-17137972 on Build GRCh38
rs767746924 Chr.22: 17138070-17138070 on Build GRCh38

View Full Product Details