Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 181004645-181004870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTNL3
Entrez Gene ID
10917
Gene Name
butyrophilin like 3
Gene Aliases
BTN9.1, BTNLR
UniGene
Hs.225949
Gene Chromosome Location
Chr.5: 180988845-181006727 on Build GRCh38
Transcripts
NM_197975.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1066893 c.846C>T p.D282D Chr.5: 181004746-181004746 on Build GRCh38
1185064 c.847G>T p.A283S Chr.5: 181004747-181004747 on Build GRCh38
1066894 c.851G>A p.R284Q Chr.5: 181004751-181004751 on Build GRCh38
4826978 c.836-1G>A p.? Chr.5: 181004735-181004735 on Build GRCh38
4155798 c.836A>G p.E279G Chr.5: 181004736-181004736 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373881919 Chr.5: 181004696-181004696 on Build GRCh38
rs561566073 Chr.5: 181004740-181004741 on Build GRCh38
rs143906320 Chr.5: 181004750-181004750 on Build GRCh38
rs780562662 Chr.5: 181004743-181004743 on Build GRCh38
rs375742795 Chr.5: 181004810-181004810 on Build GRCh38
rs768629666 Chr.5: 181004686-181004687 on Build GRCh38
rs759337826 Chr.5: 181004793-181004793 on Build GRCh38
rs548740876 Chr.5: 181004808-181004808 on Build GRCh38
rs201534771 Chr.5: 181004747-181004747 on Build GRCh38
rs777316895 Chr.5: 181004737-181004737 on Build GRCh38
rs747405558 Chr.5: 181004688-181004688 on Build GRCh38
rs202119732 Chr.5: 181004746-181004746 on Build GRCh38
rs765357378 Chr.5: 181004701-181004701 on Build GRCh38
rs200452211 Chr.5: 181004712-181004712 on Build GRCh38
rs763152483 Chr.5: 181004766-181004766 on Build GRCh38
rs151034827 Chr.5: 181004720-181004720 on Build GRCh38
rs772391437 Chr.5: 181004694-181004695 on Build GRCh38
rs371013216 Chr.5: 181004711-181004711 on Build GRCh38
rs1017557264 Chr.5: 181004797-181004797 on Build GRCh38
rs772617373 Chr.5: 181004687-181004687 on Build GRCh38
rs187604443 Chr.5: 181004770-181004770 on Build GRCh38
rs893254603 Chr.5: 181004761-181004761 on Build GRCh38
rs755788330 Chr.5: 181004726-181004726 on Build GRCh38
rs774442663 Chr.5: 181004783-181004783 on Build GRCh38
rs1036285276 Chr.5: 181004682-181004682 on Build GRCh38
rs375992218 Chr.5: 181004697-181004697 on Build GRCh38
rs371454208 Chr.5: 181004758-181004758 on Build GRCh38
rs536706389 Chr.5: 181004677-181004677 on Build GRCh38
rs201134313 Chr.5: 181004811-181004811 on Build GRCh38
rs573867040 Chr.5: 181004725-181004725 on Build GRCh38
rs545554746 Chr.5: 181004705-181004705 on Build GRCh38
rs72494582 Chr.5: 181004765-181004765 on Build GRCh38
rs748392659 Chr.5: 181004751-181004751 on Build GRCh38
rs758863164 Chr.5: 181004742-181004742 on Build GRCh38
rs754839312 Chr.5: 181004719-181004719 on Build GRCh38
rs757859738 Chr.5: 181004821-181004821 on Build GRCh38
rs372382578 Chr.5: 181004699-181004699 on Build GRCh38
rs371953707 Chr.5: 181004683-181004683 on Build GRCh38
rs767337962 Chr.5: 181004799-181004799 on Build GRCh38
rs748792154 Chr.5: 181004740-181004740 on Build GRCh38
rs773226201 Chr.5: 181004704-181004704 on Build GRCh38

View Full Product Details