Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 44358110-44358369 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCAIM
Entrez Gene ID
285343
Gene Name
T-cell activation inhibitor, mitochondrial
Gene Aliases
C3orf23, TOAG-1, TOAG1
UniGene
Hs.55131
Gene Chromosome Location
Chr.3: 44338119-44409448 on Build GRCh38
Transcripts
NM_001029839.2 NM_001029840.2 NM_001282913.1 NM_001282914.1 NM_001282915.1 NM_173826.3 XM_017006251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
297858 c.202T>G p.F68V Chr.3: 44358253-44358253 on Build GRCh38
4957804 c.181A>T p.S61C Chr.3: 44358232-44358232 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs187426580 Chr.3: 44358330-44358330 on Build GRCh38
rs754827682 Chr.3: 44358196-44358196 on Build GRCh38
rs1041176976 Chr.3: 44358286-44358286 on Build GRCh38
rs866565144 Chr.3: 44358242-44358242 on Build GRCh38
rs201221161 Chr.3: 44358203-44358203 on Build GRCh38
rs746343248 Chr.3: 44358192-44358193 on Build GRCh38
rs776310217 Chr.3: 44358202-44358203 on Build GRCh38
rs200460046 Chr.3: 44358201-44358201 on Build GRCh38
rs911469992 Chr.3: 44358274-44358274 on Build GRCh38
rs749644218 Chr.3: 44358175-44358176 on Build GRCh38
rs755305562 Chr.3: 44358289-44358289 on Build GRCh38
rs375210033 Chr.3: 44358328-44358328 on Build GRCh38
rs796837433 Chr.3: 44358202-44358203 on Build GRCh38
rs751135348 Chr.3: 44358190-44358190 on Build GRCh38
rs372867357 Chr.3: 44358331-44358331 on Build GRCh38
rs747140795 Chr.3: 44358270-44358270 on Build GRCh38
rs777132485 Chr.3: 44358291-44358291 on Build GRCh38
rs775803103 Chr.3: 44358266-44358266 on Build GRCh38
rs748147911 Chr.3: 44358227-44358227 on Build GRCh38
rs200366497 Chr.3: 44358284-44358284 on Build GRCh38
rs766027450 Chr.3: 44358178-44358178 on Build GRCh38
rs199853182 Chr.3: 44358225-44358225 on Build GRCh38
rs4682970 Chr.3: 44358176-44358176 on Build GRCh38
rs779340322 Chr.3: 44358191-44358192 on Build GRCh38
rs771276388 Chr.3: 44358181-44358183 on Build GRCh38
rs771637344 Chr.3: 44358192-44358195 on Build GRCh38
rs571181884 Chr.3: 44358264-44358264 on Build GRCh38
rs756029086 Chr.3: 44358230-44358230 on Build GRCh38
rs1054821254 Chr.3: 44358158-44358158 on Build GRCh38
rs760124398 Chr.3: 44358201-44358202 on Build GRCh38
rs762468409 Chr.3: 44358293-44358293 on Build GRCh38
rs772892356 Chr.3: 44358296-44358296 on Build GRCh38
rs776420817 Chr.3: 44358329-44358329 on Build GRCh38
rs765048722 Chr.3: 44358310-44358310 on Build GRCh38
rs200065952 Chr.3: 44358208-44358208 on Build GRCh38
rs774972229 Chr.3: 44358202-44358202 on Build GRCh38
rs765817383 Chr.3: 44358294-44358294 on Build GRCh38
rs772005327 Chr.3: 44358261-44358261 on Build GRCh38
rs779010122 Chr.3: 44358240-44358240 on Build GRCh38
rs745916564 Chr.3: 44358241-44358241 on Build GRCh38
rs972784605 Chr.3: 44358174-44358174 on Build GRCh38
rs374543933 Chr.3: 44358166-44358166 on Build GRCh38

View Full Product Details