Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218400086-218400317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTDSP1
Entrez Gene ID
58190
Gene Name
CTD small phosphatase 1
Gene Aliases
NIF3, NLI-IF, NLIIF, SCP1
UniGene
Hs.444468
Gene Chromosome Location
Chr.2: 218398338-218405941 on Build GRCh38
Transcripts
NM_001206878.1 NM_021198.2 NM_182642.2 XM_017004617.1 XM_017004618.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6382065 c.6C>A p.D2E Chr.2: 218400096-218400096 on Build GRCh38
1614359 c.17_18insCA p.I7fs*104 Chr.2: 218400107-218400108 on Build GRCh38
5708073 c.67+1G>T p.? Chr.2: 218400158-218400158 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs906675880 Chr.2: 218400135-218400135 on Build GRCh38
rs919485493 Chr.2: 218400109-218400109 on Build GRCh38
rs879677750 Chr.2: 218400208-218400208 on Build GRCh38
rs1001911108 Chr.2: 218400154-218400154 on Build GRCh38
rs1012429643 Chr.2: 218400118-218400118 on Build GRCh38
rs1029507243 Chr.2: 218400248-218400248 on Build GRCh38
rs768119984 Chr.2: 218400155-218400155 on Build GRCh38
rs748796724 Chr.2: 218400131-218400131 on Build GRCh38
rs997902910 Chr.2: 218400217-218400217 on Build GRCh38
rs745478840 Chr.2: 218400120-218400120 on Build GRCh38
rs953858327 Chr.2: 218400260-218400260 on Build GRCh38
rs773593761 Chr.2: 218400168-218400168 on Build GRCh38
rs893505654 Chr.2: 218400206-218400206 on Build GRCh38
rs772437283 Chr.2: 218400166-218400166 on Build GRCh38
rs12620498 Chr.2: 218400262-218400262 on Build GRCh38
rs771005069 Chr.2: 218400160-218400160 on Build GRCh38
rs866328917 Chr.2: 218400132-218400132 on Build GRCh38
rs867989709 Chr.2: 218400172-218400172 on Build GRCh38
rs555872843 Chr.2: 218400125-218400125 on Build GRCh38
rs1010973882 Chr.2: 218400211-218400211 on Build GRCh38
rs769541036 Chr.2: 218400122-218400122 on Build GRCh38
rs577208564 Chr.2: 218400230-218400230 on Build GRCh38
rs12620519 Chr.2: 218400144-218400144 on Build GRCh38
rs537523584 Chr.2: 218400123-218400123 on Build GRCh38
rs910629366 Chr.2: 218400269-218400269 on Build GRCh38
rs749482555 Chr.2: 218400177-218400178 on Build GRCh38
rs780862243 Chr.2: 218400105-218400105 on Build GRCh38
rs986251159 Chr.2: 218400267-218400267 on Build GRCh38
rs770050335 Chr.2: 218400171-218400174 on Build GRCh38
rs558120517 Chr.2: 218400278-218400278 on Build GRCh38
rs1054799880 Chr.2: 218400189-218400189 on Build GRCh38
rs780044082 Chr.2: 218400159-218400160 on Build GRCh38
rs754114536 Chr.2: 218400259-218400259 on Build GRCh38
rs774858271 Chr.2: 218400163-218400163 on Build GRCh38
rs773537084 Chr.2: 218400176-218400177 on Build GRCh38
rs963434812 Chr.2: 218400295-218400295 on Build GRCh38
rs897778020 Chr.2: 218400255-218400255 on Build GRCh38
rs766562413 Chr.2: 218400180-218400180 on Build GRCh38
rs985335025 Chr.2: 218400256-218400256 on Build GRCh38
rs748226699 Chr.2: 218400164-218400165 on Build GRCh38
rs962693934 Chr.2: 218400268-218400268 on Build GRCh38
rs186986949 Chr.2: 218400240-218400240 on Build GRCh38
rs762243089 Chr.2: 218400165-218400165 on Build GRCh38
rs868227078 Chr.2: 218400225-218400225 on Build GRCh38
rs967046517 Chr.2: 218400233-218400233 on Build GRCh38
rs761064461 Chr.2: 218400179-218400179 on Build GRCh38
rs953990363 Chr.2: 218400234-218400235 on Build GRCh38

View Full Product Details