Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63700645-63700900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFRP1
Entrez Gene ID
10139
Gene Name
ADP ribosylation factor related protein 1
Gene Aliases
ARL18, ARP, Arp1
UniGene
Hs.389277
Gene Chromosome Location
Chr.20: 63698642-63708013 on Build GRCh38
Transcripts
NM_001134758.3 NM_001267544.2 NM_001267545.2 NM_001267546.2 NM_001267547.2 NM_001267548.2 NM_001267549.2 NM_003224.5 NR_051954.2 NR_051955.2 NR_051956.2 NR_051957.2 NR_051958.2 XM_011528482.2 XM_011528483.1 XM_017027576.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5682846 c.437A>G p.D146G Chr.20: 63700683-63700683 on Build GRCh38
6602959 c.420G>A p.T140T Chr.20: 63700700-63700700 on Build GRCh38
5741954 c.431T>C p.I144T Chr.20: 63700689-63700689 on Build GRCh38
5775011 c.441C>G p.I147M Chr.20: 63700679-63700679 on Build GRCh38
6322405 c.419C>A p.T140K Chr.20: 63700701-63700701 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773806414 Chr.20: 63700830-63700830 on Build GRCh38
rs558564969 Chr.20: 63700881-63700881 on Build GRCh38
rs1010346972 Chr.20: 63700764-63700764 on Build GRCh38
rs527250698 Chr.20: 63700724-63700724 on Build GRCh38
rs751595682 Chr.20: 63700692-63700692 on Build GRCh38
rs372480202 Chr.20: 63700711-63700711 on Build GRCh38
rs750696744 Chr.20: 63700674-63700674 on Build GRCh38
rs753608123 Chr.20: 63700744-63700744 on Build GRCh38
rs111732018 Chr.20: 63700720-63700720 on Build GRCh38
rs904702685 Chr.20: 63700784-63700784 on Build GRCh38
rs780143365 Chr.20: 63700687-63700687 on Build GRCh38
rs775473470 Chr.20: 63700730-63700730 on Build GRCh38
rs781780125 Chr.20: 63700701-63700701 on Build GRCh38
rs976365953 Chr.20: 63700861-63700861 on Build GRCh38
rs35158527 Chr.20: 63700673-63700673 on Build GRCh38
rs749165004 Chr.20: 63700747-63700747 on Build GRCh38
rs1018150406 Chr.20: 63700801-63700801 on Build GRCh38
rs1052533578 Chr.20: 63700672-63700672 on Build GRCh38
rs766524046 Chr.20: 63700751-63700751 on Build GRCh38
rs183956106 Chr.20: 63700676-63700676 on Build GRCh38
rs927280745 Chr.20: 63700718-63700718 on Build GRCh38
rs756754006 Chr.20: 63700738-63700742 on Build GRCh38
rs188627542 Chr.20: 63700765-63700765 on Build GRCh38
rs375170087 Chr.20: 63700719-63700719 on Build GRCh38
rs760520926 Chr.20: 63700665-63700665 on Build GRCh38
rs758978552 Chr.20: 63700725-63700725 on Build GRCh38
rs760429080 Chr.20: 63700732-63700732 on Build GRCh38
rs770519445 Chr.20: 63700722-63700722 on Build GRCh38
rs6011038 Chr.20: 63700829-63700829 on Build GRCh38
rs748523089 Chr.20: 63700705-63700705 on Build GRCh38
rs549883479 Chr.20: 63700782-63700782 on Build GRCh38
rs972592890 Chr.20: 63700663-63700663 on Build GRCh38
rs368169075 Chr.20: 63700706-63700706 on Build GRCh38
rs747836586 Chr.20: 63700872-63700872 on Build GRCh38
rs377717326 Chr.20: 63700750-63700750 on Build GRCh38
rs771598856 Chr.20: 63700876-63700876 on Build GRCh38
rs546994989 Chr.20: 63700728-63700728 on Build GRCh38
rs755456323 Chr.20: 63700700-63700700 on Build GRCh38
rs566995899 Chr.20: 63700757-63700757 on Build GRCh38
rs752431053 Chr.20: 63700664-63700664 on Build GRCh38
rs1025373107 Chr.20: 63700865-63700865 on Build GRCh38
rs569731593 Chr.20: 63700788-63700788 on Build GRCh38
rs763924270 Chr.20: 63700738-63700738 on Build GRCh38

View Full Product Details