Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 202961146-202961381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARF
Entrez Gene ID
79800
Gene Name
calcium responsive transcription factor
Gene Aliases
ALS2CR8, NYD-SP24
UniGene
Hs.444982
Gene Chromosome Location
Chr.2: 202912218-202987856 on Build GRCh38
Transcripts
NM_001104586.2 NM_001282910.1 NM_001282911.1 NM_001282912.1 NM_001322427.1 NM_001322428.1 NM_001322429.1 NM_001322430.1 NM_024744.15 NR_136330.1 NR_136331.1 XM_005246858.3 XM_005246859.3 XM_005246864.3 XM_006712760.2 XM_011511867.2 XM_011511868.2 XM_011511869.2 XM_017004961.1 XM_017004962.1 XM_017004963.1 XM_017004964.1 XM_017004965.1 XM_017004966.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1182649 c.767C>T p.P256L Chr.2: 202961361-202961361 on Build GRCh38
6620179 c.644A>G p.K215R Chr.2: 202961238-202961238 on Build GRCh38
318660 c.656C>T p.S219L Chr.2: 202961250-202961250 on Build GRCh38
461194 c.734G>C p.W245S Chr.2: 202961328-202961328 on Build GRCh38
6446564 c.771T>C p.A257A Chr.2: 202961365-202961365 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200793099 Chr.2: 202961322-202961322 on Build GRCh38
rs989497590 Chr.2: 202961174-202961174 on Build GRCh38
rs745880868 Chr.2: 202961249-202961249 on Build GRCh38
rs755987427 Chr.2: 202961251-202961251 on Build GRCh38
rs751188778 Chr.2: 202961352-202961352 on Build GRCh38
rs757207993 Chr.2: 202961362-202961365 on Build GRCh38
rs780993137 Chr.2: 202961197-202961197 on Build GRCh38
rs555538912 Chr.2: 202961191-202961191 on Build GRCh38
rs750044578 Chr.2: 202961317-202961317 on Build GRCh38
rs199607480 Chr.2: 202961278-202961278 on Build GRCh38
rs372949207 Chr.2: 202961337-202961337 on Build GRCh38
rs748233756 Chr.2: 202961299-202961299 on Build GRCh38
rs773152646 Chr.2: 202961310-202961310 on Build GRCh38
rs777088012 Chr.2: 202961329-202961329 on Build GRCh38
rs764916102 Chr.2: 202961293-202961293 on Build GRCh38
rs759933397 Chr.2: 202961336-202961336 on Build GRCh38
rs779948077 Chr.2: 202961255-202961255 on Build GRCh38
rs751139756 Chr.2: 202961186-202961186 on Build GRCh38
rs753767587 Chr.2: 202961267-202961270 on Build GRCh38
rs371806353 Chr.2: 202961196-202961196 on Build GRCh38
rs772207372 Chr.2: 202961307-202961307 on Build GRCh38
rs369372393 Chr.2: 202961326-202961326 on Build GRCh38
rs985946476 Chr.2: 202961306-202961306 on Build GRCh38
rs375198785 Chr.2: 202961256-202961256 on Build GRCh38

View Full Product Details