Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 93956719-93956980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB2
Entrez Gene ID
51676
Gene Name
ankyrin repeat and SOCS box containing 2
Gene Aliases
ASB-2
UniGene
Hs.510327
Gene Chromosome Location
Chr.14: 93934153-93976796 on Build GRCh38
Transcripts
NM_001202429.1 NM_016150.4 XM_005267758.3 XM_011536834.2 XM_017021369.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6141360 c.151A>G p.K51E Chr.14: 93956782-93956782 on Build GRCh38
4975796 c.86C>T p.S29L Chr.14: 93956847-93956847 on Build GRCh38
6050753 c.34C>G p.L12V Chr.14: 93956899-93956899 on Build GRCh38
1740074 c.103A>G p.M35V Chr.14: 93956830-93956830 on Build GRCh38
5589750 c.96G>A p.R32R Chr.14: 93956837-93956837 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763333456 Chr.14: 93956937-93956937 on Build GRCh38
rs745904365 Chr.14: 93956843-93956843 on Build GRCh38
rs777974246 Chr.14: 93956877-93956877 on Build GRCh38
rs3765494 Chr.14: 93956948-93956948 on Build GRCh38
rs751355925 Chr.14: 93956739-93956739 on Build GRCh38
rs963004876 Chr.14: 93956772-93956772 on Build GRCh38
rs779390026 Chr.14: 93956761-93956761 on Build GRCh38
rs199867477 Chr.14: 93956952-93956952 on Build GRCh38
rs532067660 Chr.14: 93956846-93956846 on Build GRCh38
rs756416347 Chr.14: 93956957-93956957 on Build GRCh38
rs773110244 Chr.14: 93956853-93956853 on Build GRCh38
rs771758276 Chr.14: 93956849-93956849 on Build GRCh38
rs766089801 Chr.14: 93956863-93956863 on Build GRCh38
rs747181826 Chr.14: 93956834-93956834 on Build GRCh38
rs886193203 Chr.14: 93956903-93956903 on Build GRCh38
rs748013844 Chr.14: 93956909-93956909 on Build GRCh38
rs764138518 Chr.14: 93956838-93956839 on Build GRCh38
rs755889756 Chr.14: 93956808-93956808 on Build GRCh38
rs201010176 Chr.14: 93956797-93956797 on Build GRCh38
rs760499296 Chr.14: 93956924-93956924 on Build GRCh38
rs369790867 Chr.14: 93956825-93956825 on Build GRCh38
rs753962132 Chr.14: 93956839-93956839 on Build GRCh38
rs767036175 Chr.14: 93956745-93956745 on Build GRCh38
rs34860343 Chr.14: 93956766-93956766 on Build GRCh38
rs199834596 Chr.14: 93956760-93956760 on Build GRCh38
rs779967090 Chr.14: 93956958-93956958 on Build GRCh38
rs772088067 Chr.14: 93956913-93956913 on Build GRCh38
rs779714373 Chr.14: 93956748-93956748 on Build GRCh38
rs182843425 Chr.14: 93956783-93956783 on Build GRCh38
rs759333718 Chr.14: 93956926-93956926 on Build GRCh38
rs769694250 Chr.14: 93956899-93956899 on Build GRCh38
rs146480080 Chr.14: 93956839-93956839 on Build GRCh38
rs551967973 Chr.14: 93956949-93956949 on Build GRCh38
rs779242464 Chr.14: 93956819-93956819 on Build GRCh38
rs371606893 Chr.14: 93956767-93956767 on Build GRCh38
rs764620347 Chr.14: 93956941-93956941 on Build GRCh38
rs771189408 Chr.14: 93956835-93956835 on Build GRCh38
rs140394606 Chr.14: 93956765-93956765 on Build GRCh38
rs781341214 Chr.14: 93956882-93956882 on Build GRCh38
rs75796328 Chr.14: 93956925-93956925 on Build GRCh38
rs373447573 Chr.14: 93956890-93956890 on Build GRCh38
rs141309480 Chr.14: 93956878-93956878 on Build GRCh38
rs774376529 Chr.14: 93956785-93956785 on Build GRCh38
rs749800659 Chr.14: 93956753-93956753 on Build GRCh38
rs753567858 Chr.14: 93956865-93956865 on Build GRCh38
rs377378577 Chr.14: 93956943-93956943 on Build GRCh38
rs755625476 Chr.14: 93956747-93956747 on Build GRCh38
rs754567556 Chr.14: 93956814-93956814 on Build GRCh38
rs369368453 Chr.14: 93956744-93956744 on Build GRCh38
rs748422419 Chr.14: 93956762-93956762 on Build GRCh38
rs376105153 Chr.14: 93956951-93956951 on Build GRCh38
rs201432422 Chr.14: 93956775-93956775 on Build GRCh38
rs987294578 Chr.14: 93956833-93956833 on Build GRCh38
rs767356431 Chr.14: 93956795-93956795 on Build GRCh38
rs560633028 Chr.14: 93956812-93956812 on Build GRCh38
rs753264787 Chr.14: 93956822-93956822 on Build GRCh38
rs760374699 Chr.14: 93956862-93956862 on Build GRCh38
rs370020819 Chr.14: 93956868-93956868 on Build GRCh38
rs148173529 Chr.14: 93956847-93956847 on Build GRCh38
rs562436235 Chr.14: 93956900-93956900 on Build GRCh38
rs199712784 Chr.14: 93956830-93956830 on Build GRCh38
rs750111909 Chr.14: 93956746-93956746 on Build GRCh38
rs761466194 Chr.14: 93956793-93956793 on Build GRCh38
rs757645618 Chr.14: 93956880-93956880 on Build GRCh38
rs762850608 Chr.14: 93956779-93956779 on Build GRCh38
rs562355115 Chr.14: 93956876-93956876 on Build GRCh38
rs764813073 Chr.14: 93956874-93956874 on Build GRCh38
rs906755405 Chr.14: 93956850-93956850 on Build GRCh38
rs752297621 Chr.14: 93956929-93956929 on Build GRCh38
rs377036265 Chr.14: 93956844-93956844 on Build GRCh38
rs773054612 Chr.14: 93956915-93956915 on Build GRCh38
rs111908446 Chr.14: 93956875-93956875 on Build GRCh38
rs1004621785 Chr.14: 93956918-93956918 on Build GRCh38
rs369456226 Chr.14: 93956774-93956774 on Build GRCh38

View Full Product Details