Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 42375629-42375890 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKK4
Entrez Gene ID
27121
Gene Name
dickkopf WNT signaling pathway inhibitor 4
Gene Aliases
DKK-4
UniGene
Hs.159311
Gene Chromosome Location
Chr.8: 42373194-42391322 on Build GRCh38
Transcripts
NM_014420.2 XM_011544488.2 XM_017013316.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3766755 c.175G>T p.E59* Chr.8: 42375767-42375767 on Build GRCh38
1250136 c.208C>T p.R70W Chr.8: 42375734-42375734 on Build GRCh38
4677676 c.146C>T p.T49I Chr.8: 42375796-42375796 on Build GRCh38
6316717 c.167C>A p.P56H Chr.8: 42375775-42375775 on Build GRCh38
6275393 c.138C>A p.D46E Chr.8: 42375804-42375804 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368117672 Chr.8: 42375832-42375832 on Build GRCh38
rs267601928 Chr.8: 42375740-42375740 on Build GRCh38
rs547215646 Chr.8: 42375737-42375737 on Build GRCh38
rs775364820 Chr.8: 42375829-42375829 on Build GRCh38
rs768346300 Chr.8: 42375842-42375842 on Build GRCh38
rs201742431 Chr.8: 42375806-42375806 on Build GRCh38
rs868153614 Chr.8: 42375777-42375777 on Build GRCh38
rs781201886 Chr.8: 42375795-42375795 on Build GRCh38
rs768651417 Chr.8: 42375728-42375728 on Build GRCh38
rs763523246 Chr.8: 42375760-42375760 on Build GRCh38
rs978240025 Chr.8: 42375679-42375679 on Build GRCh38
rs774137854 Chr.8: 42375657-42375657 on Build GRCh38
rs1036845529 Chr.8: 42375859-42375859 on Build GRCh38
rs561931135 Chr.8: 42375712-42375712 on Build GRCh38
rs751786708 Chr.8: 42375792-42375792 on Build GRCh38
rs759515472 Chr.8: 42375659-42375661 on Build GRCh38
rs770888126 Chr.8: 42375734-42375734 on Build GRCh38
rs374249506 Chr.8: 42375772-42375772 on Build GRCh38
rs775311632 Chr.8: 42375744-42375744 on Build GRCh38
rs774054532 Chr.8: 42375739-42375739 on Build GRCh38
rs114758407 Chr.8: 42375686-42375686 on Build GRCh38
rs755171982 Chr.8: 42375795-42375795 on Build GRCh38
rs751540104 Chr.8: 42375744-42375752 on Build GRCh38
rs142986794 Chr.8: 42375719-42375719 on Build GRCh38
rs868549094 Chr.8: 42375709-42375709 on Build GRCh38
rs748227396 Chr.8: 42375807-42375807 on Build GRCh38
rs565991423 Chr.8: 42375808-42375808 on Build GRCh38
rs200941514 Chr.8: 42375733-42375733 on Build GRCh38
rs535127034 Chr.8: 42375820-42375820 on Build GRCh38
rs781267904 Chr.8: 42375729-42375729 on Build GRCh38
rs751215631 Chr.8: 42375688-42375688 on Build GRCh38
rs762225766 Chr.8: 42375863-42375864 on Build GRCh38
rs753371072 Chr.8: 42375768-42375768 on Build GRCh38
rs765296632 Chr.8: 42375689-42375690 on Build GRCh38
rs925301479 Chr.8: 42375800-42375800 on Build GRCh38
rs374625940 Chr.8: 42375822-42375822 on Build GRCh38
rs779199373 Chr.8: 42375702-42375702 on Build GRCh38
rs764606744 Chr.8: 42375865-42375865 on Build GRCh38
rs758028423 Chr.8: 42375659-42375659 on Build GRCh38
rs758663008 Chr.8: 42375718-42375718 on Build GRCh38
rs747187598 Chr.8: 42375722-42375722 on Build GRCh38
rs750962176 Chr.8: 42375867-42375867 on Build GRCh38
rs111605859 Chr.8: 42375673-42375673 on Build GRCh38
rs115963393 Chr.8: 42375771-42375771 on Build GRCh38
rs771943105 Chr.8: 42375742-42375742 on Build GRCh38
rs980121502 Chr.8: 42375793-42375793 on Build GRCh38
rs891183413 Chr.8: 42375838-42375838 on Build GRCh38
rs372008071 Chr.8: 42375790-42375790 on Build GRCh38
rs779015725 Chr.8: 42375814-42375814 on Build GRCh38
rs757368338 Chr.8: 42375746-42375747 on Build GRCh38
rs767247800 Chr.8: 42375666-42375666 on Build GRCh38
rs758787588 Chr.8: 42375776-42375776 on Build GRCh38
rs754303294 Chr.8: 42375685-42375685 on Build GRCh38
rs752303033 Chr.8: 42375670-42375670 on Build GRCh38
rs200089567 Chr.8: 42375759-42375759 on Build GRCh38
rs776049231 Chr.8: 42375850-42375850 on Build GRCh38
rs146172755 Chr.8: 42375665-42375665 on Build GRCh38
rs201984841 Chr.8: 42375773-42375773 on Build GRCh38
rs761367670 Chr.8: 42375860-42375860 on Build GRCh38
rs574070198 Chr.8: 42375681-42375681 on Build GRCh38

View Full Product Details