Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 39177449-39177678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNB1
Entrez Gene ID
782
Gene Name
calcium voltage-gated channel auxiliary subunit beta 1
Gene Aliases
CAB1, CACNLB1, CCHLB1
UniGene
Hs.635
Gene Chromosome Location
Chr.17: 39173456-39197703 on Build GRCh38
Transcripts
NM_000723.4 NM_199247.2 NM_199248.2 XM_005257645.2 XM_006722072.2 XM_017025024.1 XM_017025025.1 XM_017025026.1 XM_017025027.1 XM_017025028.1 XM_017025029.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6618512 c.1205C>T p.A402V Chr.17: 39177477-39177477 on Build GRCh38
1199368 c.1153T>C p.F385L Chr.17: 39177529-39177529 on Build GRCh38
3370696 c.1224T>C p.Y408Y Chr.17: 39177458-39177458 on Build GRCh38
3370698 c.1197G>A p.E399E Chr.17: 39177485-39177485 on Build GRCh38
6618513 c.1340C>T p.A447V Chr.17: 39177477-39177477 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528722029 Chr.17: 39177549-39177549 on Build GRCh38
rs781530095 Chr.17: 39177581-39177581 on Build GRCh38
rs974750989 Chr.17: 39177583-39177583 on Build GRCh38
rs200807660 Chr.17: 39177533-39177533 on Build GRCh38
rs765732612 Chr.17: 39177555-39177555 on Build GRCh38
rs150172956 Chr.17: 39177552-39177552 on Build GRCh38
rs546971996 Chr.17: 39177611-39177611 on Build GRCh38
rs757601138 Chr.17: 39177580-39177580 on Build GRCh38
rs933992377 Chr.17: 39177599-39177599 on Build GRCh38
rs778144395 Chr.17: 39177472-39177472 on Build GRCh38
rs112991958 Chr.17: 39177536-39177536 on Build GRCh38
rs753235965 Chr.17: 39177564-39177564 on Build GRCh38
rs751853668 Chr.17: 39177578-39177578 on Build GRCh38
rs745917788 Chr.17: 39177492-39177492 on Build GRCh38
rs771166354 Chr.17: 39177476-39177476 on Build GRCh38
rs774338002 Chr.17: 39177501-39177501 on Build GRCh38
rs113530680 Chr.17: 39177537-39177537 on Build GRCh38
rs758692983 Chr.17: 39177569-39177569 on Build GRCh38
rs759016283 Chr.17: 39177548-39177548 on Build GRCh38
rs765899319 Chr.17: 39177545-39177545 on Build GRCh38
rs564195118 Chr.17: 39177651-39177651 on Build GRCh38
rs1053269839 Chr.17: 39177573-39177573 on Build GRCh38
rs892052883 Chr.17: 39177591-39177591 on Build GRCh38
rs771846387 Chr.17: 39177503-39177503 on Build GRCh38
rs768512220 Chr.17: 39177496-39177496 on Build GRCh38
rs931269932 Chr.17: 39177655-39177655 on Build GRCh38
rs764477557 Chr.17: 39177574-39177574 on Build GRCh38
rs761568285 Chr.17: 39177502-39177502 on Build GRCh38
rs72556379 Chr.17: 39177470-39177470 on Build GRCh38
rs781370846 Chr.17: 39177487-39177487 on Build GRCh38
rs772858368 Chr.17: 39177530-39177530 on Build GRCh38

View Full Product Details