Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85764074-85764305 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATOH8
Entrez Gene ID
84913
Gene Name
atonal bHLH transcription factor 8
Gene Aliases
HATH6, bHLHa21
UniGene
Hs.135569
Gene Chromosome Location
Chr.2: 85753786-85791383 on Build GRCh38
Transcripts
NM_032827.6 XM_006712122.3 XM_011533139.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5964404 c.858T>A p.A286A Chr.2: 85764080-85764080 on Build GRCh38
197519 c.943C>T p.R315C Chr.2: 85764165-85764165 on Build GRCh38
4563148 c.960G>A p.K320K Chr.2: 85764182-85764182 on Build GRCh38
6606945 c.944G>A p.R315H Chr.2: 85764166-85764166 on Build GRCh38
1615181 c.876C>T p.A292A Chr.2: 85764098-85764098 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918861105 Chr.2: 85764281-85764281 on Build GRCh38
rs377138428 Chr.2: 85764221-85764221 on Build GRCh38
rs560136015 Chr.2: 85764110-85764110 on Build GRCh38
rs777773100 Chr.2: 85764178-85764178 on Build GRCh38
rs780536843 Chr.2: 85764233-85764233 on Build GRCh38
rs773893750 Chr.2: 85764153-85764153 on Build GRCh38
rs780040578 Chr.2: 85764158-85764158 on Build GRCh38
rs757554782 Chr.2: 85764223-85764223 on Build GRCh38
rs375639850 Chr.2: 85764140-85764140 on Build GRCh38
rs746254177 Chr.2: 85764215-85764215 on Build GRCh38
rs143074049 Chr.2: 85764156-85764156 on Build GRCh38
rs972948079 Chr.2: 85764253-85764253 on Build GRCh38
rs369738451 Chr.2: 85764149-85764149 on Build GRCh38
rs111910117 Chr.2: 85764122-85764122 on Build GRCh38
rs530928877 Chr.2: 85764259-85764259 on Build GRCh38
rs934789772 Chr.2: 85764216-85764216 on Build GRCh38
rs762558662 Chr.2: 85764147-85764147 on Build GRCh38
rs766333168 Chr.2: 85764166-85764166 on Build GRCh38
rs112575919 Chr.2: 85764283-85764283 on Build GRCh38
rs771168034 Chr.2: 85764123-85764123 on Build GRCh38
rs752788861 Chr.2: 85764098-85764098 on Build GRCh38
rs757421034 Chr.2: 85764203-85764203 on Build GRCh38
rs754920791 Chr.2: 85764177-85764177 on Build GRCh38
rs375874153 Chr.2: 85764148-85764148 on Build GRCh38
rs200636678 Chr.2: 85764145-85764145 on Build GRCh38
rs962886225 Chr.2: 85764240-85764240 on Build GRCh38
rs372705499 Chr.2: 85764136-85764136 on Build GRCh38
rs563665020 Chr.2: 85764257-85764257 on Build GRCh38
rs145075676 Chr.2: 85764202-85764202 on Build GRCh38
rs760406327 Chr.2: 85764165-85764165 on Build GRCh38
rs947097690 Chr.2: 85764258-85764258 on Build GRCh38
rs988537605 Chr.2: 85764234-85764234 on Build GRCh38
rs372989892 Chr.2: 85764162-85764162 on Build GRCh38
rs374950997 Chr.2: 85764170-85764170 on Build GRCh38
rs375184554 Chr.2: 85764204-85764204 on Build GRCh38
rs751650499 Chr.2: 85764196-85764196 on Build GRCh38

View Full Product Details